Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66966133-66966406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES3
Entrez Gene ID
23491
Gene Name
carboxylesterase 3
Gene Aliases
ES31
UniGene
Hs.268700
Gene Chromosome Location
Chr.16: 66961229-66975152 on Build GRCh38
Transcripts
NM_001185177.1 NM_024922.5 XM_011522975.2 XM_011522976.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354053 c.852C>T p.S284S Chr.16: 66966276-66966276 on Build GRCh38
7259213 c.842G>A p.C281Y Chr.16: 66966266-66966266 on Build GRCh38
3510886 c.889G>A p.E297K Chr.16: 66966313-66966313 on Build GRCh38
3510885 c.820-1G>A p.? Chr.16: 66966243-66966243 on Build GRCh38
2994027 c.857C>T p.P286L Chr.16: 66966281-66966281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776592005 Chr.16: 66966325-66966325 on Build GRCh38
rs774170586 Chr.16: 66966346-66966346 on Build GRCh38
rs769002369 Chr.16: 66966219-66966219 on Build GRCh38
rs748640076 Chr.16: 66966231-66966231 on Build GRCh38
rs779298538 Chr.16: 66966290-66966290 on Build GRCh38
rs536559559 Chr.16: 66966360-66966360 on Build GRCh38
rs748502371 Chr.16: 66966309-66966309 on Build GRCh38
rs994960075 Chr.16: 66966185-66966185 on Build GRCh38
rs959668948 Chr.16: 66966213-66966213 on Build GRCh38
rs779160467 Chr.16: 66966226-66966226 on Build GRCh38
rs751337753 Chr.16: 66966260-66966260 on Build GRCh38
rs769946796 Chr.16: 66966345-66966345 on Build GRCh38
rs753319460 Chr.16: 66966371-66966371 on Build GRCh38
rs796664402 Chr.16: 66966335-66966335 on Build GRCh38
rs757160436 Chr.16: 66966193-66966193 on Build GRCh38
rs139755080 Chr.16: 66966279-66966279 on Build GRCh38
rs773396058 Chr.16: 66966242-66966242 on Build GRCh38
rs754629690 Chr.16: 66966373-66966373 on Build GRCh38
rs778481293 Chr.16: 66966374-66966374 on Build GRCh38
rs759759162 Chr.16: 66966336-66966336 on Build GRCh38
rs772488980 Chr.16: 66966239-66966239 on Build GRCh38
rs767488457 Chr.16: 66966267-66966267 on Build GRCh38
rs778488685 Chr.16: 66966314-66966314 on Build GRCh38
rs779742558 Chr.16: 66966206-66966206 on Build GRCh38
rs749924366 Chr.16: 66966274-66966274 on Build GRCh38
rs772691880 Chr.16: 66966312-66966312 on Build GRCh38
rs67674179 Chr.16: 66966364-66966364 on Build GRCh38
rs745420150 Chr.16: 66966198-66966198 on Build GRCh38
rs897920784 Chr.16: 66966174-66966174 on Build GRCh38
rs201000109 Chr.16: 66966264-66966264 on Build GRCh38
rs531786588 Chr.16: 66966161-66966161 on Build GRCh38
rs749614303 Chr.16: 66966215-66966215 on Build GRCh38
rs766727486 Chr.16: 66966343-66966343 on Build GRCh38
rs567629196 Chr.16: 66966281-66966281 on Build GRCh38
rs760764647 Chr.16: 66966207-66966207 on Build GRCh38
rs944708549 Chr.16: 66966164-66966164 on Build GRCh38
rs771272599 Chr.16: 66966247-66966247 on Build GRCh38
rs755744633 Chr.16: 66966204-66966204 on Build GRCh38
rs760893859 Chr.16: 66966245-66966245 on Build GRCh38
rs201368885 Chr.16: 66966250-66966250 on Build GRCh38
rs149807823 Chr.16: 66966282-66966282 on Build GRCh38
rs569114265 Chr.16: 66966160-66966160 on Build GRCh38
rs551889655 Chr.16: 66966249-66966249 on Build GRCh38
rs568099725 Chr.16: 66966192-66966192 on Build GRCh38
rs907086688 Chr.16: 66966253-66966253 on Build GRCh38
rs370983435 Chr.16: 66966316-66966316 on Build GRCh38
rs905388033 Chr.16: 66966194-66966194 on Build GRCh38
rs1036420228 Chr.16: 66966169-66966169 on Build GRCh38
rs71647898 Chr.16: 66966258-66966258 on Build GRCh38
rs753100869 Chr.16: 66966376-66966376 on Build GRCh38
rs755691171 Chr.16: 66966278-66966278 on Build GRCh38
rs780638499 Chr.16: 66966197-66966197 on Build GRCh38
rs376216934 Chr.16: 66966365-66966365 on Build GRCh38
rs767441063 Chr.16: 66966356-66966356 on Build GRCh38
rs145692472 Chr.16: 66966317-66966317 on Build GRCh38

View Full Product Details