Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132679669-132679920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4
Entrez Gene ID
3565
Gene Name
interleukin 4
Gene Aliases
BCGF-1, BCGF1, BSF-1, BSF1, IL-4
UniGene
Hs.73917
Gene Chromosome Location
Chr.5: 132673986-132682678 on Build GRCh38
Transcripts
NM_000589.3 NM_172348.2 NR_134248.1

Gene Symbol
LOC105379176
Entrez Gene ID
105379176
Gene Name
uncharacterized LOC105379176
Gene Aliases
-
UniGene
Hs.73917
Gene Chromosome Location
Chr.5: 132679085-132682629 on Build GRCh38
Transcripts
NM_000589.3 NM_172348.2 NR_134248.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469920 c.208T>G p.C70G Chr.5: 132679738-132679738 on Build GRCh38
6469921 c.208T>C p.C70R Chr.5: 132679738-132679738 on Build GRCh38
6229150 c.230G>A p.R77Q Chr.5: 132679760-132679760 on Build GRCh38
281909 c.262C>T p.R88C Chr.5: 132679792-132679792 on Build GRCh38
6365800 c.352G>T p.A118S Chr.5: 132679882-132679882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767406138 Chr.5: 132679868-132679868 on Build GRCh38
rs776595794 Chr.5: 132679749-132679749 on Build GRCh38
rs201694257 Chr.5: 132679751-132679751 on Build GRCh38
rs201837621 Chr.5: 132679860-132679860 on Build GRCh38
rs938917376 Chr.5: 132679776-132679776 on Build GRCh38
rs755666354 Chr.5: 132679803-132679803 on Build GRCh38
rs200667292 Chr.5: 132679712-132679712 on Build GRCh38
rs763790153 Chr.5: 132679773-132679773 on Build GRCh38
rs79908535 Chr.5: 132679856-132679856 on Build GRCh38
rs777163397 Chr.5: 132679845-132679845 on Build GRCh38
rs71645915 Chr.5: 132679889-132679889 on Build GRCh38
rs201446269 Chr.5: 132679830-132679830 on Build GRCh38
rs750376744 Chr.5: 132679870-132679870 on Build GRCh38
rs777575672 Chr.5: 132679777-132679779 on Build GRCh38
rs373334025 Chr.5: 132679855-132679855 on Build GRCh38
rs747348861 Chr.5: 132679835-132679835 on Build GRCh38
rs149950065 Chr.5: 132679883-132679883 on Build GRCh38
rs35648164 Chr.5: 132679863-132679863 on Build GRCh38
rs758244348 Chr.5: 132679771-132679771 on Build GRCh38
rs539263445 Chr.5: 132679844-132679844 on Build GRCh38
rs199819530 Chr.5: 132679822-132679822 on Build GRCh38
rs1017506989 Chr.5: 132679898-132679898 on Build GRCh38
rs200433079 Chr.5: 132679880-132679880 on Build GRCh38
rs199591873 Chr.5: 132679720-132679720 on Build GRCh38
rs200172265 Chr.5: 132679785-132679785 on Build GRCh38
rs775753738 Chr.5: 132679858-132679858 on Build GRCh38
rs757675990 Chr.5: 132679708-132679708 on Build GRCh38
rs754610167 Chr.5: 132679884-132679884 on Build GRCh38
rs1047696325 Chr.5: 132679721-132679721 on Build GRCh38
rs757168472 Chr.5: 132679792-132679792 on Build GRCh38
rs752893119 Chr.5: 132679770-132679770 on Build GRCh38
rs199936687 Chr.5: 132679833-132679833 on Build GRCh38
rs771654026 Chr.5: 132679734-132679736 on Build GRCh38
rs746758811 Chr.5: 132679781-132679783 on Build GRCh38
rs201493772 Chr.5: 132679813-132679813 on Build GRCh38
rs372754769 Chr.5: 132679690-132679690 on Build GRCh38
rs781148776 Chr.5: 132679793-132679793 on Build GRCh38
rs202231191 Chr.5: 132679867-132679867 on Build GRCh38
rs377297730 Chr.5: 132679794-132679794 on Build GRCh38
rs755754084 Chr.5: 132679876-132679876 on Build GRCh38
rs779716790 Chr.5: 132679877-132679877 on Build GRCh38
rs143377308 Chr.5: 132679806-132679806 on Build GRCh38
rs922569136 Chr.5: 132679800-132679800 on Build GRCh38
rs146713238 Chr.5: 132679861-132679861 on Build GRCh38
rs781616317 Chr.5: 132679767-132679767 on Build GRCh38
rs377174102 Chr.5: 132679779-132679779 on Build GRCh38
rs202011365 Chr.5: 132679748-132679748 on Build GRCh38
rs779912651 Chr.5: 132679805-132679805 on Build GRCh38
rs759572366 Chr.5: 132679760-132679760 on Build GRCh38
rs1050455886 Chr.5: 132679824-132679824 on Build GRCh38
rs1011277356 Chr.5: 132679810-132679810 on Build GRCh38

View Full Product Details