Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12995839-12996070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
UniGene
Hs.434232
Gene Chromosome Location
Chr.17: 12991612-13018064 on Build GRCh38
Transcripts
NM_001165962.1 NM_018127.6 NM_173717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378082 c.1690C>T p.R564C Chr.17: 12995948-12995948 on Build GRCh38
3958028 c.1687G>T p.E563* Chr.17: 12995951-12995951 on Build GRCh38
1743246 c.1692C>T p.R564R Chr.17: 12995946-12995946 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761972024 Chr.17: 12995934-12995934 on Build GRCh38
rs747926772 Chr.17: 12995893-12995893 on Build GRCh38
rs910420350 Chr.17: 12995935-12995935 on Build GRCh38
rs975428986 Chr.17: 12995965-12995965 on Build GRCh38
rs762239395 Chr.17: 12995921-12995921 on Build GRCh38
rs1000349460 Chr.17: 12995987-12995987 on Build GRCh38
rs539975783 Chr.17: 12996043-12996043 on Build GRCh38
rs755286781 Chr.17: 12995951-12995951 on Build GRCh38
rs1012640664 Chr.17: 12995873-12995873 on Build GRCh38
rs752535379 Chr.17: 12995953-12995953 on Build GRCh38
rs962053201 Chr.17: 12996033-12996033 on Build GRCh38
rs117582762 Chr.17: 12995946-12995946 on Build GRCh38
rs748818277 Chr.17: 12995904-12995904 on Build GRCh38
rs1041964996 Chr.17: 12995963-12995963 on Build GRCh38
rs376766610 Chr.17: 12995952-12995952 on Build GRCh38
rs751744788 Chr.17: 12995949-12995949 on Build GRCh38
rs369124217 Chr.17: 12995915-12995915 on Build GRCh38
rs773809126 Chr.17: 12995922-12995922 on Build GRCh38
rs370011511 Chr.17: 12995945-12995945 on Build GRCh38
rs531643962 Chr.17: 12995888-12995890 on Build GRCh38
rs981037678 Chr.17: 12995887-12995887 on Build GRCh38
rs889196932 Chr.17: 12996001-12996001 on Build GRCh38
rs920039553 Chr.17: 12995969-12995969 on Build GRCh38
rs1021388532 Chr.17: 12995874-12995874 on Build GRCh38
rs951847484 Chr.17: 12995886-12995886 on Build GRCh38
rs993527438 Chr.17: 12996044-12996044 on Build GRCh38
rs556844011 Chr.17: 12995911-12995911 on Build GRCh38
rs376288494 Chr.17: 12995885-12995885 on Build GRCh38
rs557422538 Chr.17: 12996028-12996028 on Build GRCh38
rs535705148 Chr.17: 12995871-12995871 on Build GRCh38
rs938952056 Chr.17: 12996040-12996040 on Build GRCh38
rs920398703 Chr.17: 12995876-12995876 on Build GRCh38
rs755851333 Chr.17: 12996000-12996000 on Build GRCh38
rs931276557 Chr.17: 12995983-12995983 on Build GRCh38
rs769791604 Chr.17: 12995895-12995895 on Build GRCh38
rs777715538 Chr.17: 12996003-12996003 on Build GRCh38
rs777199929 Chr.17: 12995918-12995918 on Build GRCh38
rs1018935678 Chr.17: 12996032-12996032 on Build GRCh38
rs759166244 Chr.17: 12995916-12995916 on Build GRCh38
rs1051069132 Chr.17: 12995996-12995996 on Build GRCh38
rs2286337 Chr.17: 12995902-12995902 on Build GRCh38
rs904810480 Chr.17: 12995968-12995968 on Build GRCh38
rs377346238 Chr.17: 12995917-12995917 on Build GRCh38
rs371341302 Chr.17: 12996031-12996031 on Build GRCh38
rs548590974 Chr.17: 12996019-12996019 on Build GRCh38

View Full Product Details