Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49634565-49634798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF2
Entrez Gene ID
55172
Gene Name
dynein axonemal assembly factor 2
Gene Aliases
C14orf104, CILD10, KTU, PF13
UniGene
Hs.231761
Gene Chromosome Location
Chr.14: 49625174-49635230 on Build GRCh38
Transcripts
NM_001083908.1 NM_018139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306262 c.421G>A p.V141I Chr.14: 49634729-49634729 on Build GRCh38
1147296 c.391T>A p.Y131N Chr.14: 49634759-49634759 on Build GRCh38
1147294 c.579G>A p.K193K Chr.14: 49634571-49634571 on Build GRCh38
1369918 c.399delG p.R134fs*45 Chr.14: 49634751-49634751 on Build GRCh38
7078187 c.528C>A p.F176L Chr.14: 49634622-49634622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751601404 Chr.14: 49634654-49634654 on Build GRCh38
rs770511447 Chr.14: 49634610-49634610 on Build GRCh38
rs529083217 Chr.14: 49634589-49634589 on Build GRCh38
rs772707582 Chr.14: 49634608-49634608 on Build GRCh38
rs756604353 Chr.14: 49634765-49634765 on Build GRCh38
rs372399039 Chr.14: 49634620-49634620 on Build GRCh38
rs887292354 Chr.14: 49634636-49634636 on Build GRCh38
rs572101185 Chr.14: 49634732-49634732 on Build GRCh38
rs1014750801 Chr.14: 49634772-49634772 on Build GRCh38
rs1047195978 Chr.14: 49634632-49634632 on Build GRCh38
rs1002627627 Chr.14: 49634775-49634775 on Build GRCh38
rs1008254325 Chr.14: 49634643-49634643 on Build GRCh38
rs749018309 Chr.14: 49634609-49634609 on Build GRCh38
rs755109468 Chr.14: 49634655-49634655 on Build GRCh38
rs767450283 Chr.14: 49634673-49634674 on Build GRCh38
rs745566638 Chr.14: 49634774-49634774 on Build GRCh38
rs758606806 Chr.14: 49634638-49634638 on Build GRCh38
rs759118009 Chr.14: 49634612-49634612 on Build GRCh38
rs575918888 Chr.14: 49634707-49634707 on Build GRCh38
rs745354571 Chr.14: 49634718-49634718 on Build GRCh38
rs770210221 Chr.14: 49634596-49634596 on Build GRCh38
rs759580456 Chr.14: 49634746-49634746 on Build GRCh38
rs541765275 Chr.14: 49634726-49634726 on Build GRCh38
rs1014698490 Chr.14: 49634616-49634616 on Build GRCh38
rs773867321 Chr.14: 49634611-49634611 on Build GRCh38
rs996890379 Chr.14: 49634658-49634658 on Build GRCh38
rs756448105 Chr.14: 49634678-49634678 on Build GRCh38
rs748691768 Chr.14: 49634669-49634669 on Build GRCh38
rs761527467 Chr.14: 49634734-49634734 on Build GRCh38
rs373251065 Chr.14: 49634727-49634727 on Build GRCh38
rs754170049 Chr.14: 49634770-49634770 on Build GRCh38
rs900049390 Chr.14: 49634738-49634738 on Build GRCh38
rs771875922 Chr.14: 49634776-49634776 on Build GRCh38
rs1048364248 Chr.14: 49634756-49634756 on Build GRCh38
rs886050528 Chr.14: 49634679-49634679 on Build GRCh38
rs886050529 Chr.14: 49634763-49634763 on Build GRCh38
rs771603303 Chr.14: 49634773-49634773 on Build GRCh38
rs541232616 Chr.14: 49634739-49634739 on Build GRCh38
rs200484042 Chr.14: 49634766-49634766 on Build GRCh38
rs753601740 Chr.14: 49634631-49634631 on Build GRCh38
rs748486341 Chr.14: 49634607-49634607 on Build GRCh38
rs937010655 Chr.14: 49634663-49634663 on Build GRCh38
rs778406709 Chr.14: 49634661-49634661 on Build GRCh38
rs767631402 Chr.14: 49634757-49634757 on Build GRCh38
rs2985685 Chr.14: 49634652-49634652 on Build GRCh38
rs767502889 Chr.14: 49634618-49634618 on Build GRCh38
rs749642262 Chr.14: 49634688-49634688 on Build GRCh38
rs776421647 Chr.14: 49634733-49634733 on Build GRCh38
rs985044750 Chr.14: 49634600-49634600 on Build GRCh38
rs775624264 Chr.14: 49634619-49634619 on Build GRCh38
rs375385938 Chr.14: 49634629-49634629 on Build GRCh38
rs770546577 Chr.14: 49634689-49634689 on Build GRCh38
rs1004283663 Chr.14: 49634768-49634768 on Build GRCh38
rs752795172 Chr.14: 49634762-49634762 on Build GRCh38
rs991131552 Chr.14: 49634679-49634679 on Build GRCh38

View Full Product Details