Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57439392-57439617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGNL1
Entrez Gene ID
84952
Gene Name
cingulin like 1
Gene Aliases
JACOP, PCING
UniGene
Hs.148989
Gene Chromosome Location
Chr.15: 57376487-57550727 on Build GRCh38
Transcripts
NM_001252335.1 NM_032866.4 XM_005254726.3 XM_005254727.4 XM_011522120.1 XM_011522121.1 XM_017022685.1 XM_017022686.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229367 c.1526G>A p.R509Q Chr.15: 57439525-57439525 on Build GRCh38
4399831 c.1472G>A p.S491N Chr.15: 57439471-57439471 on Build GRCh38
6402602 c.1579T>C p.F527L Chr.15: 57439578-57439578 on Build GRCh38
5849432 c.1483G>A p.E495K Chr.15: 57439482-57439482 on Build GRCh38
374188 c.1504G>T p.A502S Chr.15: 57439503-57439503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868713227 Chr.15: 57439446-57439446 on Build GRCh38
rs771924925 Chr.15: 57439425-57439425 on Build GRCh38
rs1043273569 Chr.15: 57439553-57439553 on Build GRCh38
rs749425088 Chr.15: 57439561-57439561 on Build GRCh38
rs767977447 Chr.15: 57439513-57439513 on Build GRCh38
rs751791509 Chr.15: 57439476-57439478 on Build GRCh38
rs763750155 Chr.15: 57439525-57439525 on Build GRCh38
rs745688534 Chr.15: 57439424-57439424 on Build GRCh38
rs762929584 Chr.15: 57439434-57439434 on Build GRCh38
rs754757776 Chr.15: 57439557-57439557 on Build GRCh38
rs570101289 Chr.15: 57439439-57439439 on Build GRCh38
rs371311448 Chr.15: 57439435-57439435 on Build GRCh38
rs774608330 Chr.15: 57439453-57439453 on Build GRCh38
rs745751238 Chr.15: 57439463-57439463 on Build GRCh38
rs768861679 Chr.15: 57439584-57439584 on Build GRCh38
rs538506830 Chr.15: 57439427-57439427 on Build GRCh38
rs145838565 Chr.15: 57439429-57439429 on Build GRCh38
rs144501542 Chr.15: 57439458-57439458 on Build GRCh38
rs138409850 Chr.15: 57439432-57439432 on Build GRCh38
rs770199558 Chr.15: 57439489-57439489 on Build GRCh38
rs140648360 Chr.15: 57439520-57439520 on Build GRCh38
rs377706066 Chr.15: 57439422-57439422 on Build GRCh38
rs147416502 Chr.15: 57439562-57439562 on Build GRCh38
rs755799852 Chr.15: 57439451-57439451 on Build GRCh38
rs759979646 Chr.15: 57439440-57439440 on Build GRCh38
rs749393928 Chr.15: 57439493-57439493 on Build GRCh38
rs369266028 Chr.15: 57439560-57439560 on Build GRCh38
rs753036039 Chr.15: 57439450-57439450 on Build GRCh38
rs745940993 Chr.15: 57439569-57439569 on Build GRCh38
rs761414840 Chr.15: 57439586-57439586 on Build GRCh38
rs141642509 Chr.15: 57439482-57439482 on Build GRCh38
rs753444724 Chr.15: 57439456-57439456 on Build GRCh38
rs760368009 Chr.15: 57439510-57439512 on Build GRCh38
rs762039273 Chr.15: 57439568-57439568 on Build GRCh38
rs147135049 Chr.15: 57439498-57439498 on Build GRCh38
rs756822083 Chr.15: 57439527-57439527 on Build GRCh38
rs770955649 Chr.15: 57439571-57439571 on Build GRCh38
rs770963676 Chr.15: 57439423-57439423 on Build GRCh38
rs758390323 Chr.15: 57439469-57439469 on Build GRCh38
rs773254702 Chr.15: 57439491-57439491 on Build GRCh38
rs777022302 Chr.15: 57439426-57439426 on Build GRCh38
rs1280396 Chr.15: 57439530-57439530 on Build GRCh38
rs761009584 Chr.15: 57439523-57439523 on Build GRCh38
rs774946602 Chr.15: 57439502-57439502 on Build GRCh38
rs377479463 Chr.15: 57439541-57439541 on Build GRCh38
rs368066063 Chr.15: 57439503-57439503 on Build GRCh38
rs865963737 Chr.15: 57439488-57439488 on Build GRCh38
rs752203507 Chr.15: 57439491-57439491 on Build GRCh38
rs761031948 Chr.15: 57439577-57439577 on Build GRCh38
rs982837808 Chr.15: 57439442-57439442 on Build GRCh38
rs375194804 Chr.15: 57439540-57439540 on Build GRCh38
rs759962123 Chr.15: 57439452-57439452 on Build GRCh38
rs776367087 Chr.15: 57439585-57439585 on Build GRCh38
rs149602673 Chr.15: 57439445-57439445 on Build GRCh38
rs371790651 Chr.15: 57439509-57439509 on Build GRCh38
rs147998969 Chr.15: 57439471-57439471 on Build GRCh38

View Full Product Details