Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63601181-63601410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63596225-63608898 on Build GRCh38
Transcripts
NM_203373.2 XM_005254320.3 XM_006720478.1 XM_011521468.2 XM_011521469.1 XM_011521472.2 XM_017022086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971493 c.389G>T p.G130V Chr.15: 63601349-63601349 on Build GRCh38
3711718 c.419C>T p.P140L Chr.15: 63601379-63601379 on Build GRCh38
6661953 c.424_425insG p.G144fs*3 Chr.15: 63601384-63601385 on Build GRCh38
3502967 c.390G>A p.G130G Chr.15: 63601350-63601350 on Build GRCh38
4056113 c.434C>A p.P145H Chr.15: 63601394-63601394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769984227 Chr.15: 63601304-63601304 on Build GRCh38
rs761407354 Chr.15: 63601352-63601352 on Build GRCh38
rs763572934 Chr.15: 63601347-63601347 on Build GRCh38
rs752132033 Chr.15: 63601291-63601291 on Build GRCh38
rs749492990 Chr.15: 63601308-63601308 on Build GRCh38
rs753475999 Chr.15: 63601283-63601283 on Build GRCh38
rs745829470 Chr.15: 63601334-63601334 on Build GRCh38
rs758809929 Chr.15: 63601287-63601287 on Build GRCh38
rs1021818551 Chr.15: 63601227-63601227 on Build GRCh38
rs916372850 Chr.15: 63601309-63601309 on Build GRCh38
rs368196308 Chr.15: 63601363-63601363 on Build GRCh38
rs776304424 Chr.15: 63601349-63601349 on Build GRCh38
rs755383918 Chr.15: 63601292-63601292 on Build GRCh38
rs760320732 Chr.15: 63601257-63601257 on Build GRCh38
rs371849245 Chr.15: 63601305-63601305 on Build GRCh38
rs781336583 Chr.15: 63601294-63601294 on Build GRCh38
rs994946034 Chr.15: 63601381-63601381 on Build GRCh38
rs766782034 Chr.15: 63601356-63601356 on Build GRCh38
rs760234925 Chr.15: 63601346-63601346 on Build GRCh38
rs746499898 Chr.15: 63601300-63601301 on Build GRCh38
rs770794061 Chr.15: 63601313-63601313 on Build GRCh38
rs748306299 Chr.15: 63601297-63601297 on Build GRCh38
rs751806326 Chr.15: 63601359-63601359 on Build GRCh38
rs767074316 Chr.15: 63601245-63601245 on Build GRCh38
rs773964896 Chr.15: 63601317-63601317 on Build GRCh38
rs780249092 Chr.15: 63601290-63601290 on Build GRCh38
rs967627591 Chr.15: 63601244-63601244 on Build GRCh38
rs376856221 Chr.15: 63601341-63601341 on Build GRCh38
rs545686131 Chr.15: 63601261-63601261 on Build GRCh38
rs939070232 Chr.15: 63601278-63601278 on Build GRCh38
rs558022835 Chr.15: 63601387-63601387 on Build GRCh38
rs753236869 Chr.15: 63601367-63601367 on Build GRCh38

View Full Product Details