Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17579341-17579612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT1
Entrez Gene ID
79709
Gene Name
collagen beta(1-O)galactosyltransferase 1
Gene Aliases
GLT25D1
UniGene
Hs.418795
Gene Chromosome Location
Chr.19: 17555594-17583162 on Build GRCh38
Transcripts
NM_024656.3 XM_005260080.4 XM_011528297.2 XM_011528298.2 XM_011528299.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5949119 c.1267-1_1268delGGT p.? Chr.19: 17579481-17579483 on Build GRCh38
4979753 c.1381G>A p.D461N Chr.19: 17579596-17579596 on Build GRCh38
5981489 c.1366G>A p.E456K Chr.19: 17579581-17579581 on Build GRCh38
5004671 c.1370G>A p.R457Q Chr.19: 17579585-17579585 on Build GRCh38
5056358 c.1373A>G p.E458G Chr.19: 17579588-17579588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534990847 Chr.19: 17579399-17579399 on Build GRCh38
rs766057963 Chr.19: 17579571-17579571 on Build GRCh38
rs569782772 Chr.19: 17579438-17579438 on Build GRCh38
rs764982752 Chr.19: 17579469-17579469 on Build GRCh38
rs769621751 Chr.19: 17579514-17579514 on Build GRCh38
rs774477102 Chr.19: 17579555-17579555 on Build GRCh38
rs763046306 Chr.19: 17579478-17579478 on Build GRCh38
rs200591748 Chr.19: 17579464-17579464 on Build GRCh38
rs923408975 Chr.19: 17579552-17579552 on Build GRCh38
rs572639404 Chr.19: 17579572-17579572 on Build GRCh38
rs747263236 Chr.19: 17579513-17579513 on Build GRCh38
rs914818334 Chr.19: 17579539-17579539 on Build GRCh38
rs971208400 Chr.19: 17579393-17579393 on Build GRCh38
rs1029340543 Chr.19: 17579377-17579377 on Build GRCh38
rs544877640 Chr.19: 17579483-17579483 on Build GRCh38
rs778026007 Chr.19: 17579507-17579507 on Build GRCh38
rs777777218 Chr.19: 17579591-17579591 on Build GRCh38
rs372582985 Chr.19: 17579452-17579452 on Build GRCh38
rs145353960 Chr.19: 17579570-17579570 on Build GRCh38
rs750076793 Chr.19: 17579569-17579569 on Build GRCh38
rs554683985 Chr.19: 17579444-17579444 on Build GRCh38
rs575790491 Chr.19: 17579475-17579475 on Build GRCh38
rs775094058 Chr.19: 17579531-17579531 on Build GRCh38
rs780576356 Chr.19: 17579440-17579440 on Build GRCh38
rs139673867 Chr.19: 17579585-17579585 on Build GRCh38
rs1039204810 Chr.19: 17579557-17579557 on Build GRCh38
rs891705934 Chr.19: 17579380-17579380 on Build GRCh38
rs1001216217 Chr.19: 17579411-17579411 on Build GRCh38
rs560699305 Chr.19: 17579494-17579494 on Build GRCh38
rs772646042 Chr.19: 17579491-17579492 on Build GRCh38
rs374939716 Chr.19: 17579482-17579482 on Build GRCh38
rs141915331 Chr.19: 17579459-17579459 on Build GRCh38
rs748229813 Chr.19: 17579453-17579453 on Build GRCh38
rs761428179 Chr.19: 17579484-17579484 on Build GRCh38
rs989045992 Chr.19: 17579510-17579510 on Build GRCh38
rs552600556 Chr.19: 17579388-17579388 on Build GRCh38
rs759354485 Chr.19: 17579465-17579465 on Build GRCh38
rs962890213 Chr.19: 17579443-17579443 on Build GRCh38
rs374190124 Chr.19: 17579419-17579419 on Build GRCh38
rs760169326 Chr.19: 17579540-17579542 on Build GRCh38
rs748178640 Chr.19: 17579493-17579493 on Build GRCh38
rs750079479 Chr.19: 17579486-17579486 on Build GRCh38
rs147915782 Chr.19: 17579499-17579499 on Build GRCh38
rs766999283 Chr.19: 17579566-17579566 on Build GRCh38
rs145212081 Chr.19: 17579485-17579485 on Build GRCh38
rs541154290 Chr.19: 17579492-17579492 on Build GRCh38
rs114538284 Chr.19: 17579389-17579389 on Build GRCh38
rs984860257 Chr.19: 17579451-17579451 on Build GRCh38
rs947577811 Chr.19: 17579554-17579554 on Build GRCh38
rs139965300 Chr.19: 17579565-17579565 on Build GRCh38
rs754214674 Chr.19: 17579491-17579491 on Build GRCh38
rs956016450 Chr.19: 17579508-17579508 on Build GRCh38
rs574529523 Chr.19: 17579449-17579449 on Build GRCh38
rs756480377 Chr.19: 17579431-17579431 on Build GRCh38
rs372871690 Chr.19: 17579542-17579542 on Build GRCh38
rs150637824 Chr.19: 17579490-17579490 on Build GRCh38
rs550488314 Chr.19: 17579551-17579551 on Build GRCh38
rs768662193 Chr.19: 17579548-17579548 on Build GRCh38
rs1015499536 Chr.19: 17579407-17579407 on Build GRCh38
rs771773083 Chr.19: 17579438-17579439 on Build GRCh38
rs745798315 Chr.19: 17579581-17579581 on Build GRCh38

View Full Product Details