Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 42779365-42779626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC152
Entrez Gene ID
100129792
Gene Name
coiled-coil domain containing 152
Gene Aliases
CH5400
UniGene
Hs.745017
Gene Chromosome Location
Chr.5: 42756818-42802437 on Build GRCh38
Transcripts
NM_001134848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401206 c.266A>T p.E89V Chr.5: 42779461-42779461 on Build GRCh38
5624586 c.267A>G p.E89E Chr.5: 42779462-42779462 on Build GRCh38
6513037 c.291T>A p.A97A Chr.5: 42779486-42779486 on Build GRCh38
5995798 c.263G>A p.G88D Chr.5: 42779458-42779458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531787553 Chr.5: 42779413-42779413 on Build GRCh38
rs768037239 Chr.5: 42779438-42779438 on Build GRCh38
rs772563109 Chr.5: 42779423-42779423 on Build GRCh38
rs11743506 Chr.5: 42779588-42779588 on Build GRCh38
rs189564185 Chr.5: 42779543-42779543 on Build GRCh38
rs529312636 Chr.5: 42779585-42779585 on Build GRCh38
rs890711897 Chr.5: 42779452-42779452 on Build GRCh38
rs929457659 Chr.5: 42779430-42779430 on Build GRCh38
rs756747130 Chr.5: 42779474-42779474 on Build GRCh38
rs397710812 Chr.5: 42779593-42779594 on Build GRCh38
rs762102229 Chr.5: 42779426-42779426 on Build GRCh38
rs11427149 Chr.5: 42779584-42779585 on Build GRCh38
rs527614302 Chr.5: 42779585-42779585 on Build GRCh38
rs148101297 Chr.5: 42779451-42779451 on Build GRCh38
rs1030664367 Chr.5: 42779483-42779483 on Build GRCh38
rs1048015516 Chr.5: 42779443-42779443 on Build GRCh38
rs370026878 Chr.5: 42779458-42779458 on Build GRCh38
rs989267951 Chr.5: 42779575-42779575 on Build GRCh38
rs1017576161 Chr.5: 42779459-42779459 on Build GRCh38
rs773633775 Chr.5: 42779494-42779494 on Build GRCh38
rs185663534 Chr.5: 42779527-42779527 on Build GRCh38
rs560895015 Chr.5: 42779585-42779586 on Build GRCh38

View Full Product Details