Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44398262-44398489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH8
Entrez Gene ID
23131
Gene Name
G-patch domain containing 8
Gene Aliases
GPATC8, KIAA0553
UniGene
Hs.463129
Gene Chromosome Location
Chr.17: 44395277-44503602 on Build GRCh38
Transcripts
NM_001002909.3 NM_001304939.1 NM_001304940.1 NM_001304941.1 NM_001304942.1 NM_001304943.1 XM_011524557.1 XM_011524558.1 XM_011524559.2 XM_011524561.2 XM_017024372.1 XM_017024373.1 XM_017024374.1 XM_017024375.1 XM_017024376.1 XM_017024377.1 XM_017024378.1 XM_017024379.1 XM_017024380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518079 c.3732C>T p.L1244L Chr.17: 44398345-44398345 on Build GRCh38
7264301 c.3698A>G p.Y1233C Chr.17: 44398379-44398379 on Build GRCh38
4615591 c.3706delG p.D1236fs*37 Chr.17: 44398371-44398371 on Build GRCh38
6673080 c.3735C>T p.P1245P Chr.17: 44398342-44398342 on Build GRCh38
5785779 c.3777C>T p.S1259S Chr.17: 44398300-44398300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769664637 Chr.17: 44398456-44398456 on Build GRCh38
rs964899517 Chr.17: 44398459-44398459 on Build GRCh38
rs765468740 Chr.17: 44398384-44398384 on Build GRCh38
rs138409533 Chr.17: 44398342-44398342 on Build GRCh38
rs369214287 Chr.17: 44398303-44398303 on Build GRCh38
rs775521436 Chr.17: 44398362-44398362 on Build GRCh38
rs746030826 Chr.17: 44398448-44398448 on Build GRCh38
rs757217899 Chr.17: 44398405-44398405 on Build GRCh38
rs758943008 Chr.17: 44398388-44398388 on Build GRCh38
rs781491789 Chr.17: 44398412-44398412 on Build GRCh38
rs142995489 Chr.17: 44398370-44398370 on Build GRCh38
rs138768525 Chr.17: 44398293-44398293 on Build GRCh38
rs778231356 Chr.17: 44398391-44398391 on Build GRCh38
rs987697969 Chr.17: 44398425-44398425 on Build GRCh38
rs889673589 Chr.17: 44398445-44398445 on Build GRCh38
rs955039028 Chr.17: 44398395-44398395 on Build GRCh38
rs201037319 Chr.17: 44398400-44398400 on Build GRCh38
rs755380635 Chr.17: 44398378-44398378 on Build GRCh38
rs779475597 Chr.17: 44398308-44398308 on Build GRCh38
rs566521603 Chr.17: 44398387-44398387 on Build GRCh38
rs947203747 Chr.17: 44398372-44398372 on Build GRCh38
rs755289846 Chr.17: 44398307-44398307 on Build GRCh38
rs146140295 Chr.17: 44398464-44398464 on Build GRCh38
rs748593696 Chr.17: 44398314-44398314 on Build GRCh38
rs776714240 Chr.17: 44398345-44398345 on Build GRCh38
rs762946542 Chr.17: 44398365-44398365 on Build GRCh38
rs749973691 Chr.17: 44398374-44398374 on Build GRCh38
rs747059872 Chr.17: 44398340-44398340 on Build GRCh38
rs780037640 Chr.17: 44398458-44398458 on Build GRCh38
rs762257124 Chr.17: 44398342-44398342 on Build GRCh38
rs769419684 Chr.17: 44398358-44398358 on Build GRCh38
rs928325298 Chr.17: 44398360-44398360 on Build GRCh38
rs758494011 Chr.17: 44398331-44398331 on Build GRCh38
rs145721422 Chr.17: 44398337-44398337 on Build GRCh38
rs745680526 Chr.17: 44398349-44398349 on Build GRCh38
rs764094874 Chr.17: 44398366-44398366 on Build GRCh38
rs768776392 Chr.17: 44398465-44398465 on Build GRCh38
rs760339 Chr.17: 44398302-44398302 on Build GRCh38
rs1018194736 Chr.17: 44398440-44398440 on Build GRCh38
rs1006809745 Chr.17: 44398457-44398457 on Build GRCh38
rs757014684 Chr.17: 44398292-44398292 on Build GRCh38
rs774057588 Chr.17: 44398368-44398368 on Build GRCh38

View Full Product Details