Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20117355-20117628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR4K17
Entrez Gene ID
390436
Gene Name
olfactory receptor family 4 subfamily K member 17
Gene Aliases
OR14-29
UniGene
Hs.553765
Gene Chromosome Location
Chr.14: 20117407-20118438 on Build GRCh38
Transcripts
NM_001004715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3936445 c.208G>C p.V70L Chr.14: 20117614-20117614 on Build GRCh38
5955670 c.206C>T p.T69I Chr.14: 20117612-20117612 on Build GRCh38
1368778 c.48C>A p.F16L Chr.14: 20117454-20117454 on Build GRCh38
1368777 c.16T>G p.S6A Chr.14: 20117422-20117422 on Build GRCh38
6851023 c.15T>C p.F5F Chr.14: 20117421-20117421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749656343 Chr.14: 20117563-20117563 on Build GRCh38
rs750203683 Chr.14: 20117537-20117537 on Build GRCh38
rs767858497 Chr.14: 20117382-20117382 on Build GRCh38
rs762238496 Chr.14: 20117488-20117488 on Build GRCh38
rs745675692 Chr.14: 20117467-20117467 on Build GRCh38
rs767095346 Chr.14: 20117437-20117437 on Build GRCh38
rs989537523 Chr.14: 20117410-20117410 on Build GRCh38
rs536593108 Chr.14: 20117409-20117409 on Build GRCh38
rs760368508 Chr.14: 20117572-20117572 on Build GRCh38
rs759186612 Chr.14: 20117434-20117434 on Build GRCh38
rs778813559 Chr.14: 20117465-20117465 on Build GRCh38
rs376010796 Chr.14: 20117552-20117552 on Build GRCh38
rs767162452 Chr.14: 20117502-20117502 on Build GRCh38
rs150087607 Chr.14: 20117594-20117594 on Build GRCh38
rs757214759 Chr.14: 20117521-20117521 on Build GRCh38
rs763273256 Chr.14: 20117496-20117496 on Build GRCh38
rs138358295 Chr.14: 20117595-20117595 on Build GRCh38
rs762150658 Chr.14: 20117414-20117414 on Build GRCh38
rs755197204 Chr.14: 20117550-20117550 on Build GRCh38
rs780310715 Chr.14: 20117547-20117547 on Build GRCh38
rs747107709 Chr.14: 20117479-20117479 on Build GRCh38
rs747140954 Chr.14: 20117548-20117548 on Build GRCh38
rs751402331 Chr.14: 20117596-20117596 on Build GRCh38
rs763863864 Chr.14: 20117581-20117581 on Build GRCh38
rs141335641 Chr.14: 20117444-20117444 on Build GRCh38
rs774786932 Chr.14: 20117564-20117564 on Build GRCh38
rs896923827 Chr.14: 20117481-20117481 on Build GRCh38
rs770314899 Chr.14: 20117493-20117493 on Build GRCh38
rs368048415 Chr.14: 20117408-20117408 on Build GRCh38
rs374774223 Chr.14: 20117494-20117494 on Build GRCh38
rs750788527 Chr.14: 20117433-20117433 on Build GRCh38
rs200403018 Chr.14: 20117388-20117388 on Build GRCh38
rs773629673 Chr.14: 20117562-20117562 on Build GRCh38
rs770911165 Chr.14: 20117387-20117387 on Build GRCh38
rs145178521 Chr.14: 20117517-20117517 on Build GRCh38
rs924403768 Chr.14: 20117405-20117405 on Build GRCh38
rs776703913 Chr.14: 20117483-20117483 on Build GRCh38
rs372021676 Chr.14: 20117558-20117558 on Build GRCh38
rs572811388 Chr.14: 20117561-20117561 on Build GRCh38
rs772218078 Chr.14: 20117393-20117393 on Build GRCh38
rs776262554 Chr.14: 20117585-20117585 on Build GRCh38
rs148373137 Chr.14: 20117530-20117530 on Build GRCh38
rs78711883 Chr.14: 20117413-20117413 on Build GRCh38
rs201465638 Chr.14: 20117516-20117516 on Build GRCh38
rs752226373 Chr.14: 20117439-20117439 on Build GRCh38
rs1031523764 Chr.14: 20117576-20117576 on Build GRCh38
rs866244331 Chr.14: 20117454-20117454 on Build GRCh38
rs61743111 Chr.14: 20117425-20117425 on Build GRCh38
rs758194242 Chr.14: 20117545-20117545 on Build GRCh38
rs761450836 Chr.14: 20117592-20117592 on Build GRCh38
rs772316895 Chr.14: 20117468-20117468 on Build GRCh38
rs140042697 Chr.14: 20117515-20117515 on Build GRCh38
rs200610810 Chr.14: 20117471-20117471 on Build GRCh38
rs562025396 Chr.14: 20117411-20117411 on Build GRCh38
rs536068350 Chr.14: 20117480-20117480 on Build GRCh38
rs757269705 Chr.14: 20117464-20117464 on Build GRCh38
rs764145049 Chr.14: 20117603-20117603 on Build GRCh38
rs922083600 Chr.14: 20117389-20117389 on Build GRCh38
rs369744365 Chr.14: 20117556-20117556 on Build GRCh38
rs755615271 Chr.14: 20117440-20117440 on Build GRCh38
rs764878924 Chr.14: 20117605-20117605 on Build GRCh38
rs766818757 Chr.14: 20117447-20117447 on Build GRCh38
rs367810132 Chr.14: 20117513-20117513 on Build GRCh38
rs371242633 Chr.14: 20117491-20117491 on Build GRCh38
rs949771513 Chr.14: 20117509-20117509 on Build GRCh38
rs753432742 Chr.14: 20117519-20117519 on Build GRCh38

View Full Product Details