Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5287762-5287995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC29A4
Entrez Gene ID
222962
Gene Name
solute carrier family 29 member 4
Gene Aliases
ENT4, PMAT
UniGene
Hs.4302
Gene Chromosome Location
Chr.7: 5274311-5304073 on Build GRCh38
Transcripts
NM_001040661.1 NM_001300847.1 NM_153247.2 XM_005249658.4 XM_006715667.3 XM_006715668.2 XM_011515200.2 XM_011515201.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4162294 c.39C>A p.S13R Chr.7: 5287855-5287855 on Build GRCh38
6781271 c.22C>T p.R8C Chr.7: 5287838-5287838 on Build GRCh38
5766161 c.117G>A p.A39A Chr.7: 5287933-5287933 on Build GRCh38
3881727 c.162G>A p.T54T Chr.7: 5287978-5287978 on Build GRCh38
6364037 c.41T>C p.V14A Chr.7: 5287857-5287857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143921693 Chr.7: 5287932-5287932 on Build GRCh38
rs770079019 Chr.7: 5287826-5287826 on Build GRCh38
rs760138659 Chr.7: 5287911-5287911 on Build GRCh38
rs77039967 Chr.7: 5287933-5287933 on Build GRCh38
rs777288431 Chr.7: 5287918-5287926 on Build GRCh38
rs200574190 Chr.7: 5287923-5287923 on Build GRCh38
rs762356263 Chr.7: 5287842-5287842 on Build GRCh38
rs772702987 Chr.7: 5287855-5287855 on Build GRCh38
rs778675832 Chr.7: 5287810-5287810 on Build GRCh38
rs559185534 Chr.7: 5287872-5287872 on Build GRCh38
rs775032578 Chr.7: 5287970-5287970 on Build GRCh38
rs779006549 Chr.7: 5287936-5287936 on Build GRCh38
rs770552581 Chr.7: 5287935-5287948 on Build GRCh38
rs753559891 Chr.7: 5287867-5287867 on Build GRCh38
rs752725965 Chr.7: 5287785-5287786 on Build GRCh38
rs781411514 Chr.7: 5287817-5287817 on Build GRCh38
rs765958145 Chr.7: 5287864-5287864 on Build GRCh38
rs73332823 Chr.7: 5287886-5287886 on Build GRCh38
rs780157571 Chr.7: 5287930-5287930 on Build GRCh38
rs1053821207 Chr.7: 5287875-5287875 on Build GRCh38
rs755223089 Chr.7: 5287935-5287935 on Build GRCh38
rs541082501 Chr.7: 5287839-5287839 on Build GRCh38
rs780440606 Chr.7: 5287830-5287830 on Build GRCh38
rs982243270 Chr.7: 5287962-5287962 on Build GRCh38
rs147508642 Chr.7: 5287966-5287966 on Build GRCh38
rs372332816 Chr.7: 5287853-5287853 on Build GRCh38
rs143263478 Chr.7: 5287964-5287964 on Build GRCh38
rs375388445 Chr.7: 5287876-5287876 on Build GRCh38
rs554717558 Chr.7: 5287789-5287789 on Build GRCh38
rs749562136 Chr.7: 5287831-5287831 on Build GRCh38
rs748090137 Chr.7: 5287811-5287811 on Build GRCh38
rs760181725 Chr.7: 5287856-5287856 on Build GRCh38
rs573375566 Chr.7: 5287825-5287825 on Build GRCh38
rs746533860 Chr.7: 5287933-5287938 on Build GRCh38
rs752990749 Chr.7: 5287907-5287907 on Build GRCh38
rs137876561 Chr.7: 5287926-5287926 on Build GRCh38
rs1019490656 Chr.7: 5287921-5287921 on Build GRCh38
rs367596044 Chr.7: 5287787-5287787 on Build GRCh38
rs779321095 Chr.7: 5287901-5287901 on Build GRCh38
rs140965974 Chr.7: 5287900-5287900 on Build GRCh38
rs952602380 Chr.7: 5287802-5287802 on Build GRCh38
rs776922614 Chr.7: 5287796-5287796 on Build GRCh38
rs749983424 Chr.7: 5287792-5287792 on Build GRCh38
rs777919702 Chr.7: 5287938-5287951 on Build GRCh38
rs777860087 Chr.7: 5287879-5287879 on Build GRCh38
rs542964211 Chr.7: 5287909-5287909 on Build GRCh38
rs780379301 Chr.7: 5287887-5287887 on Build GRCh38
rs748454436 Chr.7: 5287949-5287949 on Build GRCh38
rs774615892 Chr.7: 5287841-5287841 on Build GRCh38
rs200818994 Chr.7: 5287955-5287955 on Build GRCh38
rs770396590 Chr.7: 5287913-5287913 on Build GRCh38
rs145276221 Chr.7: 5287902-5287902 on Build GRCh38
rs759152269 Chr.7: 5287869-5287869 on Build GRCh38
rs750476897 Chr.7: 5287880-5287880 on Build GRCh38
rs753562546 Chr.7: 5287806-5287806 on Build GRCh38
rs149488369 Chr.7: 5287927-5287927 on Build GRCh38
rs758318760 Chr.7: 5287816-5287816 on Build GRCh38
rs758783624 Chr.7: 5287954-5287954 on Build GRCh38
rs776055320 Chr.7: 5287915-5287915 on Build GRCh38
rs747493694 Chr.7: 5287960-5287960 on Build GRCh38
rs771487953 Chr.7: 5287963-5287963 on Build GRCh38
rs769433504 Chr.7: 5287968-5287968 on Build GRCh38
rs772500738 Chr.7: 5287903-5287903 on Build GRCh38
rs199555796 Chr.7: 5287924-5287924 on Build GRCh38
rs755748607 Chr.7: 5287794-5287794 on Build GRCh38
rs766087136 Chr.7: 5287800-5287800 on Build GRCh38
rs754825751 Chr.7: 5287807-5287807 on Build GRCh38
rs764447381 Chr.7: 5287784-5287784 on Build GRCh38
rs757826145 Chr.7: 5287844-5287846 on Build GRCh38

View Full Product Details