Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113428313-113428566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDSL
Entrez Gene ID
113675
Gene Name
serine dehydratase like
Gene Aliases
SDH 2, SDS-RS1, TDH, cSDH
UniGene
Hs.337594
Gene Chromosome Location
Chr.12: 113422265-113438277 on Build GRCh38
Transcripts
NM_001304993.1 NM_138432.3 XM_005253831.4 XM_011537846.2 XM_017018763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639366 c.178G>A p.A60T Chr.12: 113428423-113428423 on Build GRCh38
691981 c.177G>A p.M59I Chr.12: 113428422-113428422 on Build GRCh38
6425139 c.207C>A p.C69* Chr.12: 113428452-113428452 on Build GRCh38
3456433 c.190T>G p.C64G Chr.12: 113428435-113428435 on Build GRCh38
4896059 c.209C>T p.S70F Chr.12: 113428454-113428454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555843256 Chr.12: 113428476-113428476 on Build GRCh38
rs1039959276 Chr.12: 113428430-113428430 on Build GRCh38
rs140649236 Chr.12: 113428394-113428394 on Build GRCh38
rs1013341690 Chr.12: 113428387-113428387 on Build GRCh38
rs544044229 Chr.12: 113428506-113428506 on Build GRCh38
rs766903045 Chr.12: 113428436-113428436 on Build GRCh38
rs900123904 Chr.12: 113428461-113428461 on Build GRCh38
rs773756087 Chr.12: 113428418-113428418 on Build GRCh38
rs916491765 Chr.12: 113428518-113428518 on Build GRCh38
rs374457448 Chr.12: 113428437-113428437 on Build GRCh38
rs781111497 Chr.12: 113428485-113428485 on Build GRCh38
rs940008740 Chr.12: 113428530-113428530 on Build GRCh38
rs959380137 Chr.12: 113428362-113428362 on Build GRCh38
rs981783982 Chr.12: 113428528-113428528 on Build GRCh38
rs762986945 Chr.12: 113428407-113428407 on Build GRCh38
rs577555864 Chr.12: 113428519-113428519 on Build GRCh38
rs769761472 Chr.12: 113428397-113428397 on Build GRCh38
rs368347918 Chr.12: 113428468-113428468 on Build GRCh38
rs1037023400 Chr.12: 113428534-113428534 on Build GRCh38
rs372442064 Chr.12: 113428470-113428470 on Build GRCh38
rs533725585 Chr.12: 113428441-113428441 on Build GRCh38
rs529329380 Chr.12: 113428359-113428359 on Build GRCh38
rs566359648 Chr.12: 113428434-113428434 on Build GRCh38
rs995733279 Chr.12: 113428350-113428350 on Build GRCh38
rs148370808 Chr.12: 113428335-113428335 on Build GRCh38
rs768786292 Chr.12: 113428386-113428386 on Build GRCh38
rs765496481 Chr.12: 113428495-113428496 on Build GRCh38
rs929736440 Chr.12: 113428351-113428351 on Build GRCh38
rs762001722 Chr.12: 113428470-113428471 on Build GRCh38
rs747615464 Chr.12: 113428339-113428339 on Build GRCh38
rs369406367 Chr.12: 113428508-113428508 on Build GRCh38
rs548878499 Chr.12: 113428457-113428457 on Build GRCh38
rs1051141488 Chr.12: 113428516-113428516 on Build GRCh38
rs4468391 Chr.12: 113428368-113428368 on Build GRCh38
rs1024613195 Chr.12: 113428388-113428388 on Build GRCh38
rs375387370 Chr.12: 113428492-113428492 on Build GRCh38
rs199965669 Chr.12: 113428393-113428393 on Build GRCh38
rs376277048 Chr.12: 113428352-113428352 on Build GRCh38
rs551852388 Chr.12: 113428404-113428404 on Build GRCh38
rs753666811 Chr.12: 113428471-113428471 on Build GRCh38
rs760304225 Chr.12: 113428438-113428438 on Build GRCh38
rs368995434 Chr.12: 113428361-113428361 on Build GRCh38
rs538401561 Chr.12: 113428520-113428520 on Build GRCh38
rs770933490 Chr.12: 113428416-113428416 on Build GRCh38
rs750722804 Chr.12: 113428443-113428443 on Build GRCh38

View Full Product Details