Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41842103-41842348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL10
Entrez Gene ID
317719
Gene Name
kelch like family member 10
Gene Aliases
SPGF11
UniGene
Hs.127510
Gene Chromosome Location
Chr.17: 41835685-41848347 on Build GRCh38
Transcripts
NM_152467.3 XM_011524706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223876 c.604G>A p.A202T Chr.17: 41842232-41842232 on Build GRCh38
1239273 c.522C>A p.V174V Chr.17: 41842150-41842150 on Build GRCh38
4753373 c.578A>G p.D193G Chr.17: 41842206-41842206 on Build GRCh38
148273 c.571G>A p.E191K Chr.17: 41842199-41842199 on Build GRCh38
979339 c.601G>T p.D201Y Chr.17: 41842229-41842229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782247845 Chr.17: 41842285-41842285 on Build GRCh38
rs782284363 Chr.17: 41842155-41842155 on Build GRCh38
rs1046079994 Chr.17: 41842214-41842214 on Build GRCh38
rs782186513 Chr.17: 41842270-41842270 on Build GRCh38
rs929428278 Chr.17: 41842153-41842153 on Build GRCh38
rs782820025 Chr.17: 41842197-41842197 on Build GRCh38
rs574533496 Chr.17: 41842174-41842174 on Build GRCh38
rs564904298 Chr.17: 41842264-41842264 on Build GRCh38
rs782610139 Chr.17: 41842279-41842279 on Build GRCh38
rs1001972869 Chr.17: 41842247-41842247 on Build GRCh38
rs371552751 Chr.17: 41842265-41842265 on Build GRCh38
rs541818300 Chr.17: 41842193-41842193 on Build GRCh38
rs781926688 Chr.17: 41842184-41842184 on Build GRCh38
rs116420871 Chr.17: 41842275-41842275 on Build GRCh38
rs1048016944 Chr.17: 41842168-41842168 on Build GRCh38
rs200286521 Chr.17: 41842137-41842137 on Build GRCh38
rs374553069 Chr.17: 41842255-41842255 on Build GRCh38
rs782786057 Chr.17: 41842263-41842263 on Build GRCh38
rs761919561 Chr.17: 41842215-41842215 on Build GRCh38
rs147108868 Chr.17: 41842222-41842222 on Build GRCh38
rs199599875 Chr.17: 41842235-41842235 on Build GRCh38
rs200355741 Chr.17: 41842144-41842144 on Build GRCh38
rs782215386 Chr.17: 41842188-41842188 on Build GRCh38
rs201438905 Chr.17: 41842250-41842250 on Build GRCh38

View Full Product Details