Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25450220-25450477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARMIL1
Entrez Gene ID
55604
Gene Name
capping protein regulator and myosin 1 linker 1
Gene Aliases
CARMIL, CARMIL1a, LRRC16, LRRC16A, dJ501N12.1, dJ501N12.5
UniGene
Hs.649550
Gene Chromosome Location
Chr.6: 25279428-25620530 on Build GRCh38
Transcripts
NM_001173977.1 NM_017640.5 XM_005249218.1 XM_017011008.1 XM_017011009.1 XM_017011010.1 XM_017011011.1 XM_017011012.1 XM_017011013.1 XM_017011014.1 XM_017011015.1 XM_017011016.1 XM_017011017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3860798 c.540+1G>A p.? Chr.6: 25450410-25450410 on Build GRCh38
1647928 c.54A>G p.Q18Q Chr.6: 25450406-25450406 on Build GRCh38
3860799 c.57+1G>A p.? Chr.6: 25450410-25450410 on Build GRCh38
3860797 c.540+1G>A p.? Chr.6: 25450410-25450410 on Build GRCh38
7143360 c.481C>G p.Q161E Chr.6: 25450350-25450350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747633709 Chr.6: 25450390-25450390 on Build GRCh38
rs758422625 Chr.6: 25450448-25450448 on Build GRCh38
rs752762409 Chr.6: 25450325-25450325 on Build GRCh38
rs972573327 Chr.6: 25450288-25450288 on Build GRCh38
rs952110902 Chr.6: 25450310-25450310 on Build GRCh38
rs766131623 Chr.6: 25450430-25450430 on Build GRCh38
rs765442409 Chr.6: 25450445-25450445 on Build GRCh38
rs778229556 Chr.6: 25450343-25450343 on Build GRCh38
rs760565249 Chr.6: 25450422-25450422 on Build GRCh38
rs765304361 Chr.6: 25450322-25450322 on Build GRCh38
rs755160993 Chr.6: 25450316-25450316 on Build GRCh38
rs771759479 Chr.6: 25450395-25450395 on Build GRCh38
rs759434525 Chr.6: 25450443-25450443 on Build GRCh38
rs766114592 Chr.6: 25450299-25450299 on Build GRCh38
rs747536663 Chr.6: 25450353-25450353 on Build GRCh38
rs757601933 Chr.6: 25450369-25450369 on Build GRCh38
rs939912306 Chr.6: 25450412-25450412 on Build GRCh38
rs747288175 Chr.6: 25450337-25450338 on Build GRCh38
rs748909814 Chr.6: 25450382-25450382 on Build GRCh38
rs1013959860 Chr.6: 25450259-25450259 on Build GRCh38
rs533023516 Chr.6: 25450431-25450431 on Build GRCh38
rs774211137 Chr.6: 25450389-25450389 on Build GRCh38
rs961249269 Chr.6: 25450264-25450264 on Build GRCh38
rs985212232 Chr.6: 25450333-25450333 on Build GRCh38
rs187145285 Chr.6: 25450380-25450380 on Build GRCh38
rs372848909 Chr.6: 25450313-25450313 on Build GRCh38
rs146409780 Chr.6: 25450260-25450260 on Build GRCh38
rs752964466 Chr.6: 25450447-25450447 on Build GRCh38
rs371033239 Chr.6: 25450330-25450330 on Build GRCh38

View Full Product Details