Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33722513-33722774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
UniGene
Hs.160324
Gene Chromosome Location
Chr.21: 33642400-33899861 on Build GRCh38
Transcripts
NM_001001132.1 NM_003024.2 XM_005261026.2 XM_005261027.1 XM_005261031.1 XM_011529691.1 XM_011529692.1 XM_011529693.2 XM_017028428.1 XM_017028429.1 XM_017028430.1 XM_017028431.1 XM_017028432.1 XM_017028433.1 XM_017028434.1 XM_017028435.1 XM_017028436.1 XM_017028438.1 XM_017028439.1 XM_017028440.1 XM_017028441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211175 c.140T>A p.F47Y Chr.21: 33722606-33722606 on Build GRCh38
6411905 c.134G>A p.R45K Chr.21: 33722600-33722600 on Build GRCh38
3550424 c.157T>A p.L53I Chr.21: 33722623-33722623 on Build GRCh38
3550422 c.157T>A p.L53I Chr.21: 33722623-33722623 on Build GRCh38
3550423 c.157T>A p.L53I Chr.21: 33722623-33722623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764512085 Chr.21: 33722655-33722655 on Build GRCh38
rs780167385 Chr.21: 33722682-33722682 on Build GRCh38
rs563383929 Chr.21: 33722594-33722594 on Build GRCh38
rs780426134 Chr.21: 33722558-33722567 on Build GRCh38
rs753128394 Chr.21: 33722542-33722547 on Build GRCh38
rs779025038 Chr.21: 33722680-33722680 on Build GRCh38
rs745906380 Chr.21: 33722681-33722681 on Build GRCh38
rs761496241 Chr.21: 33722607-33722607 on Build GRCh38
rs987248011 Chr.21: 33722708-33722708 on Build GRCh38
rs746737709 Chr.21: 33722579-33722579 on Build GRCh38
rs764465112 Chr.21: 33722544-33722544 on Build GRCh38
rs757478858 Chr.21: 33722679-33722679 on Build GRCh38
rs1028883221 Chr.21: 33722706-33722706 on Build GRCh38
rs1031173472 Chr.21: 33722668-33722668 on Build GRCh38
rs371748151 Chr.21: 33722704-33722704 on Build GRCh38
rs749510229 Chr.21: 33722545-33722545 on Build GRCh38
rs753267635 Chr.21: 33722650-33722650 on Build GRCh38
rs182707345 Chr.21: 33722742-33722742 on Build GRCh38
rs776464722 Chr.21: 33722697-33722697 on Build GRCh38
rs768487154 Chr.21: 33722585-33722585 on Build GRCh38
rs775436809 Chr.21: 33722572-33722572 on Build GRCh38
rs73368079 Chr.21: 33722544-33722544 on Build GRCh38
rs999308393 Chr.21: 33722643-33722643 on Build GRCh38
rs754190857 Chr.21: 33722669-33722669 on Build GRCh38
rs369315838 Chr.21: 33722684-33722684 on Build GRCh38
rs764858527 Chr.21: 33722541-33722542 on Build GRCh38
rs771947751 Chr.21: 33722568-33722568 on Build GRCh38
rs771784691 Chr.21: 33722560-33722567 on Build GRCh38
rs746959118 Chr.21: 33722604-33722605 on Build GRCh38
rs764851463 Chr.21: 33722615-33722615 on Build GRCh38
rs749462106 Chr.21: 33722559-33722560 on Build GRCh38
rs1018774028 Chr.21: 33722728-33722728 on Build GRCh38
rs769451989 Chr.21: 33722699-33722699 on Build GRCh38
rs762586266 Chr.21: 33722622-33722622 on Build GRCh38
rs770879789 Chr.21: 33722605-33722605 on Build GRCh38
rs768516525 Chr.21: 33722693-33722693 on Build GRCh38
rs758769207 Chr.21: 33722543-33722544 on Build GRCh38
rs745997061 Chr.21: 33722567-33722567 on Build GRCh38
rs796977287 Chr.21: 33722545-33722546 on Build GRCh38
rs768073680 Chr.21: 33722634-33722634 on Build GRCh38
rs776357486 Chr.21: 33722602-33722602 on Build GRCh38
rs890122419 Chr.21: 33722746-33722746 on Build GRCh38
rs373376953 Chr.21: 33722545-33722545 on Build GRCh38
rs759237042 Chr.21: 33722540-33722542 on Build GRCh38
rs190028287 Chr.21: 33722705-33722705 on Build GRCh38
rs752033410 Chr.21: 33722544-33722545 on Build GRCh38
rs749237118 Chr.21: 33722707-33722707 on Build GRCh38
rs533491843 Chr.21: 33722545-33722546 on Build GRCh38
rs757484392 Chr.21: 33722547-33722547 on Build GRCh38
rs948188099 Chr.21: 33722577-33722577 on Build GRCh38
rs756633876 Chr.21: 33722651-33722651 on Build GRCh38
rs779015550 Chr.21: 33722560-33722560 on Build GRCh38
rs773018547 Chr.21: 33722560-33722560 on Build GRCh38
rs140646821 Chr.21: 33722616-33722616 on Build GRCh38

View Full Product Details