Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93656352-93656613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF2
Entrez Gene ID
5253
Gene Name
PHD finger protein 2
Gene Aliases
CENP-35, GRC5, JHDM1E
UniGene
Hs.211441
Gene Chromosome Location
Chr.9: 93576627-93679587 on Build GRCh38
Transcripts
NM_005392.3 XM_005252051.2 XM_006717143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6874642 c.1061G>A p.R354K Chr.9: 93656509-93656509 on Build GRCh38
3659714 c.1092C>T p.P364P Chr.9: 93656540-93656540 on Build GRCh38
6855101 c.1074C>T p.G358G Chr.9: 93656522-93656522 on Build GRCh38
4713971 c.1106C>T p.A369V Chr.9: 93656554-93656554 on Build GRCh38
753851 c.1084C>G p.Q362E Chr.9: 93656532-93656532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746810340 Chr.9: 93656467-93656467 on Build GRCh38
rs372093036 Chr.9: 93656495-93656495 on Build GRCh38
rs1034839720 Chr.9: 93656379-93656379 on Build GRCh38
rs775499720 Chr.9: 93656465-93656465 on Build GRCh38
rs756410471 Chr.9: 93656554-93656554 on Build GRCh38
rs139496293 Chr.9: 93656593-93656593 on Build GRCh38
rs956429427 Chr.9: 93656534-93656534 on Build GRCh38
rs752005736 Chr.9: 93656509-93656509 on Build GRCh38
rs575748741 Chr.9: 93656436-93656436 on Build GRCh38
rs768285853 Chr.9: 93656380-93656380 on Build GRCh38
rs756183926 Chr.9: 93656439-93656439 on Build GRCh38
rs768446881 Chr.9: 93656582-93656582 on Build GRCh38
rs576941191 Chr.9: 93656498-93656498 on Build GRCh38
rs755358606 Chr.9: 93656518-93656518 on Build GRCh38
rs777846852 Chr.9: 93656555-93656555 on Build GRCh38
rs780158689 Chr.9: 93656572-93656572 on Build GRCh38
rs768444625 Chr.9: 93656478-93656478 on Build GRCh38
rs978899114 Chr.9: 93656525-93656525 on Build GRCh38
rs368274465 Chr.9: 93656553-93656553 on Build GRCh38
rs972816864 Chr.9: 93656474-93656474 on Build GRCh38
rs373680350 Chr.9: 93656440-93656440 on Build GRCh38
rs1037764305 Chr.9: 93656393-93656393 on Build GRCh38
rs772196725 Chr.9: 93656458-93656458 on Build GRCh38
rs764773950 Chr.9: 93656491-93656491 on Build GRCh38
rs993538171 Chr.9: 93656426-93656426 on Build GRCh38
rs776165547 Chr.9: 93656437-93656437 on Build GRCh38
rs776175669 Chr.9: 93656481-93656481 on Build GRCh38
rs746898327 Chr.9: 93656573-93656573 on Build GRCh38
rs781089470 Chr.9: 93656585-93656585 on Build GRCh38
rs375214167 Chr.9: 93656501-93656501 on Build GRCh38
rs897892353 Chr.9: 93656397-93656397 on Build GRCh38
rs761567621 Chr.9: 93656482-93656482 on Build GRCh38
rs959270487 Chr.9: 93656386-93656386 on Build GRCh38
rs990730328 Chr.9: 93656418-93656418 on Build GRCh38
rs143194843 Chr.9: 93656455-93656455 on Build GRCh38
rs1023180230 Chr.9: 93656453-93656453 on Build GRCh38
rs199681826 Chr.9: 93656570-93656570 on Build GRCh38
rs150964160 Chr.9: 93656526-93656526 on Build GRCh38

View Full Product Details