Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91055554-91055807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
UniGene
Hs.125750
Gene Chromosome Location
Chr.12: 91050491-91058354 on Build GRCh38
Transcripts
NM_007035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466355 c.617A>T p.N206I Chr.12: 91055665-91055665 on Build GRCh38
3466354 c.720C>T p.A240A Chr.12: 91055562-91055562 on Build GRCh38
1364824 c.498G>A p.E166E Chr.12: 91055784-91055784 on Build GRCh38
5913399 c.615G>T p.R205S Chr.12: 91055667-91055667 on Build GRCh38
1364823 c.577A>C p.N193H Chr.12: 91055705-91055705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139692564 Chr.12: 91055586-91055586 on Build GRCh38
rs554422120 Chr.12: 91055742-91055742 on Build GRCh38
rs780507479 Chr.12: 91055666-91055666 on Build GRCh38
rs121917860 Chr.12: 91055762-91055762 on Build GRCh38
rs539537384 Chr.12: 91055668-91055668 on Build GRCh38
rs149224582 Chr.12: 91055738-91055738 on Build GRCh38
rs752726071 Chr.12: 91055609-91055609 on Build GRCh38
rs747616158 Chr.12: 91055667-91055667 on Build GRCh38
rs578034447 Chr.12: 91055646-91055646 on Build GRCh38
rs763731124 Chr.12: 91055722-91055722 on Build GRCh38
rs866677626 Chr.12: 91055594-91055594 on Build GRCh38
rs763099091 Chr.12: 91055698-91055698 on Build GRCh38
rs755859874 Chr.12: 91055621-91055621 on Build GRCh38
rs372272138 Chr.12: 91055718-91055718 on Build GRCh38
rs778354357 Chr.12: 91055659-91055659 on Build GRCh38
rs1017125971 Chr.12: 91055749-91055749 on Build GRCh38
rs747414542 Chr.12: 91055746-91055746 on Build GRCh38
rs748564427 Chr.12: 91055758-91055758 on Build GRCh38
rs757898662 Chr.12: 91055740-91055740 on Build GRCh38
rs1035688652 Chr.12: 91055780-91055780 on Build GRCh38
rs369794921 Chr.12: 91055692-91055692 on Build GRCh38
rs947195505 Chr.12: 91055728-91055728 on Build GRCh38
rs762249896 Chr.12: 91055724-91055724 on Build GRCh38
rs766635508 Chr.12: 91055712-91055712 on Build GRCh38
rs914005658 Chr.12: 91055616-91055616 on Build GRCh38
rs756854659 Chr.12: 91055726-91055726 on Build GRCh38
rs745405837 Chr.12: 91055660-91055660 on Build GRCh38
rs753698147 Chr.12: 91055723-91055723 on Build GRCh38
rs770168873 Chr.12: 91055693-91055693 on Build GRCh38
rs779596762 Chr.12: 91055745-91055745 on Build GRCh38
rs755510675 Chr.12: 91055751-91055751 on Build GRCh38
rs773620958 Chr.12: 91055694-91055694 on Build GRCh38
rs978646549 Chr.12: 91055669-91055669 on Build GRCh38
rs756940768 Chr.12: 91055649-91055649 on Build GRCh38
rs768888284 Chr.12: 91055685-91055685 on Build GRCh38
rs121917862 Chr.12: 91055638-91055638 on Build GRCh38
rs775280323 Chr.12: 91055714-91055714 on Build GRCh38
rs769312068 Chr.12: 91055617-91055617 on Build GRCh38
rs150036305 Chr.12: 91055603-91055603 on Build GRCh38
rs781367942 Chr.12: 91055754-91055754 on Build GRCh38
rs753820015 Chr.12: 91055664-91055664 on Build GRCh38
rs942455680 Chr.12: 91055619-91055619 on Build GRCh38
rs142473139 Chr.12: 91055775-91055775 on Build GRCh38
rs777208206 Chr.12: 91055686-91055686 on Build GRCh38
rs145384644 Chr.12: 91055733-91055733 on Build GRCh38

View Full Product Details