Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21499495-21499724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL3
Entrez Gene ID
56339
Gene Name
methyltransferase like 3
Gene Aliases
IME4, M6A, MT-A70, Spo8
UniGene
Hs.168799
Gene Chromosome Location
Chr.14: 21498131-21511375 on Build GRCh38
Transcripts
NM_019852.4 XM_006720206.3 XM_011536968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4701833 c.1433A>G p.H478R Chr.14: 21499511-21499511 on Build GRCh38
1640041 c.1403G>A p.R468Q Chr.14: 21499541-21499541 on Build GRCh38
3706184 c.1392A>C p.Q464H Chr.14: 21499552-21499552 on Build GRCh38
469795 c.1441G>A p.E481K Chr.14: 21499503-21499503 on Build GRCh38
1300488 c.1357G>A p.D453N Chr.14: 21499587-21499587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946965341 Chr.14: 21499520-21499520 on Build GRCh38
rs767615017 Chr.14: 21499548-21499548 on Build GRCh38
rs749643805 Chr.14: 21499629-21499629 on Build GRCh38
rs774309591 Chr.14: 21499577-21499577 on Build GRCh38
rs772781305 Chr.14: 21499627-21499627 on Build GRCh38
rs1015449570 Chr.14: 21499657-21499657 on Build GRCh38
rs917007264 Chr.14: 21499688-21499688 on Build GRCh38
rs992617811 Chr.14: 21499667-21499667 on Build GRCh38
rs140146410 Chr.14: 21499546-21499546 on Build GRCh38
rs761801841 Chr.14: 21499591-21499591 on Build GRCh38
rs760040385 Chr.14: 21499634-21499634 on Build GRCh38
rs746954678 Chr.14: 21499602-21499602 on Build GRCh38
rs750226006 Chr.14: 21499576-21499576 on Build GRCh38
rs774880730 Chr.14: 21499633-21499633 on Build GRCh38
rs1012499313 Chr.14: 21499645-21499645 on Build GRCh38
rs540836022 Chr.14: 21499635-21499635 on Build GRCh38
rs774768803 Chr.14: 21499540-21499540 on Build GRCh38
rs984419689 Chr.14: 21499655-21499655 on Build GRCh38
rs571760146 Chr.14: 21499624-21499624 on Build GRCh38
rs779995154 Chr.14: 21499592-21499592 on Build GRCh38
rs146603540 Chr.14: 21499522-21499522 on Build GRCh38
rs772993260 Chr.14: 21499574-21499575 on Build GRCh38
rs934416154 Chr.14: 21499549-21499549 on Build GRCh38
rs952670013 Chr.14: 21499639-21499639 on Build GRCh38
rs752850121 Chr.14: 21499551-21499551 on Build GRCh38
rs576831643 Chr.14: 21499692-21499692 on Build GRCh38
rs961194038 Chr.14: 21499663-21499663 on Build GRCh38
rs201104563 Chr.14: 21499606-21499606 on Build GRCh38
rs771220600 Chr.14: 21499631-21499631 on Build GRCh38
rs577897694 Chr.14: 21499673-21499673 on Build GRCh38
rs760807140 Chr.14: 21499646-21499646 on Build GRCh38
rs373137461 Chr.14: 21499622-21499622 on Build GRCh38
rs569532008 Chr.14: 21499552-21499552 on Build GRCh38
rs149041981 Chr.14: 21499560-21499560 on Build GRCh38
rs557657730 Chr.14: 21499642-21499642 on Build GRCh38

View Full Product Details