Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169926765-169927010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD7
Entrez Gene ID
344658
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
-
UniGene
Hs.439922
Gene Chromosome Location
Chr.3: 169911572-169939175 on Build GRCh38
Transcripts
NM_001304366.1 NM_182610.3 NR_130713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3589988 c.516C>T p.H172H Chr.3: 169926778-169926778 on Build GRCh38
213357 c.734C>T p.T245M Chr.3: 169926996-169926996 on Build GRCh38
3915343 c.597G>A p.E199E Chr.3: 169926859-169926859 on Build GRCh38
6545028 c.646G>T p.V216F Chr.3: 169926908-169926908 on Build GRCh38
5681996 c.569A>T p.N190I Chr.3: 169926831-169926831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753686799 Chr.3: 169926931-169926931 on Build GRCh38
rs572528023 Chr.3: 169926847-169926847 on Build GRCh38
rs543334158 Chr.3: 169926982-169926982 on Build GRCh38
rs1045017316 Chr.3: 169926834-169926834 on Build GRCh38
rs765490784 Chr.3: 169926911-169926911 on Build GRCh38
rs764708176 Chr.3: 169926791-169926791 on Build GRCh38
rs765695055 Chr.3: 169926802-169926802 on Build GRCh38
rs563568150 Chr.3: 169926867-169926867 on Build GRCh38
rs746517989 Chr.3: 169926882-169926882 on Build GRCh38
rs964871329 Chr.3: 169926872-169926872 on Build GRCh38
rs751662393 Chr.3: 169926823-169926823 on Build GRCh38
rs10513680 Chr.3: 169926922-169926922 on Build GRCh38
rs994763320 Chr.3: 169926914-169926914 on Build GRCh38
rs142003969 Chr.3: 169926808-169926808 on Build GRCh38
rs971452506 Chr.3: 169926950-169926950 on Build GRCh38
rs201077493 Chr.3: 169926957-169926957 on Build GRCh38
rs762436083 Chr.3: 169926910-169926910 on Build GRCh38
rs746713922 Chr.3: 169926974-169926974 on Build GRCh38
rs777269122 Chr.3: 169926904-169926904 on Build GRCh38
rs371063340 Chr.3: 169926926-169926926 on Build GRCh38
rs778522262 Chr.3: 169926940-169926940 on Build GRCh38
rs761211369 Chr.3: 169926789-169926789 on Build GRCh38
rs763038409 Chr.3: 169926915-169926915 on Build GRCh38
rs779843525 Chr.3: 169926970-169926970 on Build GRCh38
rs773660579 Chr.3: 169926912-169926912 on Build GRCh38
rs766408429 Chr.3: 169926822-169926822 on Build GRCh38
rs1006175531 Chr.3: 169926857-169926857 on Build GRCh38
rs949414315 Chr.3: 169926813-169926813 on Build GRCh38
rs1052520914 Chr.3: 169926806-169926806 on Build GRCh38
rs777693047 Chr.3: 169926840-169926840 on Build GRCh38
rs115323462 Chr.3: 169926794-169926794 on Build GRCh38
rs762075412 Chr.3: 169926798-169926798 on Build GRCh38
rs751572209 Chr.3: 169926920-169926920 on Build GRCh38
rs766603276 Chr.3: 169926916-169926916 on Build GRCh38
rs755988273 Chr.3: 169926829-169926829 on Build GRCh38
rs758054843 Chr.3: 169926965-169926965 on Build GRCh38
rs749281246 Chr.3: 169926854-169926854 on Build GRCh38
rs768345557 Chr.3: 169926890-169926890 on Build GRCh38
rs745470133 Chr.3: 169926865-169926865 on Build GRCh38
rs778806992 Chr.3: 169926863-169926863 on Build GRCh38
rs757247832 Chr.3: 169926938-169926938 on Build GRCh38
rs150704593 Chr.3: 169926861-169926861 on Build GRCh38
rs775080190 Chr.3: 169926878-169926878 on Build GRCh38
rs758690200 Chr.3: 169926810-169926810 on Build GRCh38
rs750242678 Chr.3: 169926947-169926947 on Build GRCh38

View Full Product Details