Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123651843-123652116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL38
Entrez Gene ID
340359
Gene Name
kelch like family member 38
Gene Aliases
C8ORFK36
UniGene
Hs.450252
Gene Chromosome Location
Chr.8: 123644442-123653829 on Build GRCh38
Transcripts
NM_001081675.2 XM_005250901.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6696775 c.817C>G p.L273V Chr.8: 123652110-123652110 on Build GRCh38
294509 c.962G>A p.R321Q Chr.8: 123651965-123651965 on Build GRCh38
6696778 c.976T>C p.S326P Chr.8: 123651951-123651951 on Build GRCh38
6112065 c.904G>T p.V302F Chr.8: 123652023-123652023 on Build GRCh38
6112066 c.926C>A p.T309N Chr.8: 123652001-123652001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374245279 Chr.8: 123652075-123652075 on Build GRCh38
rs766585743 Chr.8: 123652034-123652034 on Build GRCh38
rs201958346 Chr.8: 123651911-123651911 on Build GRCh38
rs141903589 Chr.8: 123652077-123652077 on Build GRCh38
rs201007388 Chr.8: 123651988-123651988 on Build GRCh38
rs901340403 Chr.8: 123651930-123651930 on Build GRCh38
rs377275127 Chr.8: 123652056-123652056 on Build GRCh38
rs753521696 Chr.8: 123651916-123651917 on Build GRCh38
rs747354953 Chr.8: 123652090-123652090 on Build GRCh38
rs775022860 Chr.8: 123651938-123651938 on Build GRCh38
rs780188773 Chr.8: 123651965-123651965 on Build GRCh38
rs372000942 Chr.8: 123651933-123651933 on Build GRCh38
rs769435513 Chr.8: 123651981-123651981 on Build GRCh38
rs372092088 Chr.8: 123651964-123651964 on Build GRCh38
rs781202050 Chr.8: 123651922-123651922 on Build GRCh38
rs755523078 Chr.8: 123652005-123652005 on Build GRCh38
rs888217723 Chr.8: 123651962-123651962 on Build GRCh38
rs776379203 Chr.8: 123651888-123651888 on Build GRCh38
rs1023226446 Chr.8: 123651976-123651976 on Build GRCh38
rs762469267 Chr.8: 123651949-123651949 on Build GRCh38
rs748680546 Chr.8: 123651970-123651970 on Build GRCh38
rs752211216 Chr.8: 123651903-123651903 on Build GRCh38
rs778160654 Chr.8: 123652084-123652084 on Build GRCh38
rs754827752 Chr.8: 123651953-123651965 on Build GRCh38
rs767329808 Chr.8: 123651958-123651958 on Build GRCh38
rs375114991 Chr.8: 123651966-123651966 on Build GRCh38
rs767393891 Chr.8: 123652000-123652000 on Build GRCh38
rs768165532 Chr.8: 123651991-123651991 on Build GRCh38
rs1035870612 Chr.8: 123651935-123651935 on Build GRCh38
rs375130098 Chr.8: 123651947-123651947 on Build GRCh38
rs748486170 Chr.8: 123651940-123651940 on Build GRCh38
rs762318828 Chr.8: 123651882-123651882 on Build GRCh38
rs750210063 Chr.8: 123652023-123652023 on Build GRCh38
rs780177938 Chr.8: 123651928-123651928 on Build GRCh38
rs11780509 Chr.8: 123651890-123651890 on Build GRCh38
rs11784175 Chr.8: 123651873-123651873 on Build GRCh38
rs763977074 Chr.8: 123651950-123651950 on Build GRCh38
rs769353078 Chr.8: 123651871-123651871 on Build GRCh38
rs368547076 Chr.8: 123651932-123651932 on Build GRCh38
rs775032794 Chr.8: 123651985-123651985 on Build GRCh38
rs757071306 Chr.8: 123651914-123651914 on Build GRCh38
rs772515615 Chr.8: 123651971-123651971 on Build GRCh38
rs755141076 Chr.8: 123651969-123651969 on Build GRCh38
rs559576404 Chr.8: 123651941-123651941 on Build GRCh38
rs746113617 Chr.8: 123651923-123651923 on Build GRCh38
rs992338801 Chr.8: 123651945-123651945 on Build GRCh38
rs766064191 Chr.8: 123651910-123651910 on Build GRCh38
rs369848638 Chr.8: 123652057-123652057 on Build GRCh38
rs368159677 Chr.8: 123651954-123651954 on Build GRCh38
rs747377832 Chr.8: 123651973-123651973 on Build GRCh38
rs200699799 Chr.8: 123652024-123652024 on Build GRCh38
rs764819514 Chr.8: 123651892-123651892 on Build GRCh38
rs11784192 Chr.8: 123651927-123651927 on Build GRCh38
rs779126192 Chr.8: 123652063-123652063 on Build GRCh38
rs776092269 Chr.8: 123651942-123651942 on Build GRCh38
rs879728921 Chr.8: 123652036-123652036 on Build GRCh38
rs528393188 Chr.8: 123651961-123651961 on Build GRCh38
rs16898693 Chr.8: 123651999-123651999 on Build GRCh38
rs550689227 Chr.8: 123652002-123652002 on Build GRCh38
rs751500203 Chr.8: 123651952-123651952 on Build GRCh38
rs775038532 Chr.8: 123651948-123651948 on Build GRCh38

View Full Product Details