Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48406892-48407119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNB1
Entrez Gene ID
5364
Gene Name
plexin B1
Gene Aliases
PLEXIN-B1, PLXN5, SEP
UniGene
Hs.476209
Gene Chromosome Location
Chr.3: 48403854-48430324 on Build GRCh38
Transcripts
NM_001130082.2 NM_002673.5 XM_011533833.2 XM_011533834.1 XM_011533835.1 XM_011533836.1 XM_011533837.2 XM_017006630.1 XM_017006631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945534 c.6138G>A p.K2046K Chr.3: 48407041-48407041 on Build GRCh38
1221291 c.6095C>T p.P2032L Chr.3: 48407084-48407084 on Build GRCh38
254896 c.6118C>T p.R2040W Chr.3: 48407061-48407061 on Build GRCh38
6750683 c.6152+2T>C p.? Chr.3: 48407025-48407025 on Build GRCh38
4715947 c.6131G>A p.R2044Q Chr.3: 48407048-48407048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769584343 Chr.3: 48406983-48406983 on Build GRCh38
rs1057301399 Chr.3: 48406969-48406969 on Build GRCh38
rs377630056 Chr.3: 48406986-48406986 on Build GRCh38
rs1005405755 Chr.3: 48406958-48406958 on Build GRCh38
rs747737833 Chr.3: 48406946-48406946 on Build GRCh38
rs764116531 Chr.3: 48407007-48407007 on Build GRCh38
rs776693598 Chr.3: 48407012-48407012 on Build GRCh38
rs376211982 Chr.3: 48407056-48407056 on Build GRCh38
rs762638688 Chr.3: 48407048-48407048 on Build GRCh38
rs779829758 Chr.3: 48406922-48406922 on Build GRCh38
rs780882079 Chr.3: 48406942-48406942 on Build GRCh38
rs556501307 Chr.3: 48406992-48406992 on Build GRCh38
rs200211751 Chr.3: 48407039-48407039 on Build GRCh38
rs370249939 Chr.3: 48407061-48407061 on Build GRCh38
rs79109532 Chr.3: 48407050-48407050 on Build GRCh38
rs34596197 Chr.3: 48406995-48406996 on Build GRCh38
rs766221983 Chr.3: 48407049-48407049 on Build GRCh38
rs747019545 Chr.3: 48406923-48406923 on Build GRCh38
rs755753973 Chr.3: 48407083-48407083 on Build GRCh38
rs369432147 Chr.3: 48406991-48406991 on Build GRCh38
rs761265388 Chr.3: 48406995-48406995 on Build GRCh38
rs763503522 Chr.3: 48406924-48406924 on Build GRCh38
rs192546829 Chr.3: 48406973-48406973 on Build GRCh38
rs765428171 Chr.3: 48407020-48407020 on Build GRCh38
rs777535314 Chr.3: 48407084-48407084 on Build GRCh38
rs761961777 Chr.3: 48407013-48407013 on Build GRCh38
rs752241808 Chr.3: 48407073-48407073 on Build GRCh38
rs948803298 Chr.3: 48406990-48406990 on Build GRCh38
rs980102467 Chr.3: 48406974-48406974 on Build GRCh38

View Full Product Details