Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78104246-78104509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGK1
Entrez Gene ID
5230
Gene Name
phosphoglycerate kinase 1
Gene Aliases
HEL-S-68p, MIG10, PGKA
UniGene
Hs.78771
Gene Chromosome Location
Chr.X: 78104169-78126827 on Build GRCh38
Transcripts
NM_000291.3

SNP Detail

SNP ID Chromosome Location
rs782400066 Chr.X: 78104454-78104454 on Build GRCh38
rs782280948 Chr.X: 78104329-78104329 on Build GRCh38
rs782639483 Chr.X: 78104393-78104393 on Build GRCh38
rs372263316 Chr.X: 78104289-78104289 on Build GRCh38
rs192936390 Chr.X: 78104281-78104281 on Build GRCh38
rs979901871 Chr.X: 78104277-78104277 on Build GRCh38
rs782351595 Chr.X: 78104443-78104443 on Build GRCh38
rs782021500 Chr.X: 78104318-78104318 on Build GRCh38
rs988767849 Chr.X: 78104269-78104269 on Build GRCh38
rs781897274 Chr.X: 78104347-78104347 on Build GRCh38
rs782483245 Chr.X: 78104380-78104380 on Build GRCh38
rs781979836 Chr.X: 78104446-78104446 on Build GRCh38
rs782752618 Chr.X: 78104330-78104330 on Build GRCh38
rs927088261 Chr.X: 78104280-78104280 on Build GRCh38
rs373149332 Chr.X: 78104320-78104320 on Build GRCh38
rs782310362 Chr.X: 78104294-78104294 on Build GRCh38
rs781839448 Chr.X: 78104278-78104278 on Build GRCh38
rs782429511 Chr.X: 78104355-78104355 on Build GRCh38
rs782715366 Chr.X: 78104357-78104357 on Build GRCh38
rs907560306 Chr.X: 78104287-78104287 on Build GRCh38
rs984474789 Chr.X: 78104283-78104283 on Build GRCh38
rs375219335 Chr.X: 78104332-78104332 on Build GRCh38
rs782559909 Chr.X: 78104426-78104426 on Build GRCh38
rs782312507 Chr.X: 78104322-78104322 on Build GRCh38
rs781939036 Chr.X: 78104327-78104327 on Build GRCh38
rs202247339 Chr.X: 78104339-78104339 on Build GRCh38
rs782223094 Chr.X: 78104314-78104314 on Build GRCh38
rs112702955 Chr.X: 78104271-78104271 on Build GRCh38
rs781858405 Chr.X: 78104315-78104315 on Build GRCh38
rs782601783 Chr.X: 78104305-78104305 on Build GRCh38
rs782252282 Chr.X: 78104449-78104449 on Build GRCh38
rs971288997 Chr.X: 78104273-78104273 on Build GRCh38
rs782275817 Chr.X: 78104394-78104394 on Build GRCh38
rs782123518 Chr.X: 78104337-78104337 on Build GRCh38
rs782599913 Chr.X: 78104272-78104272 on Build GRCh38
rs781825608 Chr.X: 78104370-78104370 on Build GRCh38
rs782675431 Chr.X: 78104290-78104290 on Build GRCh38
rs11541573 Chr.X: 78104323-78104323 on Build GRCh38
rs782108964 Chr.X: 78104440-78104441 on Build GRCh38
rs782774228 Chr.X: 78104288-78104288 on Build GRCh38
rs782566582 Chr.X: 78104412-78104412 on Build GRCh38
rs782201500 Chr.X: 78104431-78104431 on Build GRCh38
rs889929794 Chr.X: 78104481-78104481 on Build GRCh38
rs782000499 Chr.X: 78104359-78104359 on Build GRCh38

View Full Product Details