Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58509792-58510059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A5
Entrez Gene ID
10998
Gene Name
solute carrier family 27 member 5
Gene Aliases
ACSB, ACSVL6, BACS, BAL, FACVL3, FATP-5, FATP5, VLACSR, VLCS-H2, VLCSH2
UniGene
Hs.292177
Gene Chromosome Location
Chr.19: 58498333-58512065 on Build GRCh38
Transcripts
NM_001321196.1 NM_012254.2 XM_011526364.1 XM_017026214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1326106 c.1043G>A p.G348D Chr.19: 58509861-58509861 on Build GRCh38
386997 c.972C>A p.A324A Chr.19: 58509932-58509932 on Build GRCh38
7210585 c.930G>A p.E310E Chr.19: 58509974-58509974 on Build GRCh38
6229450 c.1042G>A p.G348S Chr.19: 58509862-58509862 on Build GRCh38
6781182 c.960C>A p.S320S Chr.19: 58509944-58509944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772325864 Chr.19: 58509898-58509898 on Build GRCh38
rs779175109 Chr.19: 58509890-58509890 on Build GRCh38
rs370721451 Chr.19: 58510037-58510037 on Build GRCh38
rs761593254 Chr.19: 58509837-58509837 on Build GRCh38
rs370687843 Chr.19: 58509959-58509959 on Build GRCh38
rs765613329 Chr.19: 58509973-58509973 on Build GRCh38
rs748024217 Chr.19: 58510023-58510023 on Build GRCh38
rs34274259 Chr.19: 58509980-58509980 on Build GRCh38
rs772845251 Chr.19: 58509964-58509964 on Build GRCh38
rs753097332 Chr.19: 58509978-58509978 on Build GRCh38
rs930579932 Chr.19: 58510021-58510021 on Build GRCh38
rs201282504 Chr.19: 58509861-58509861 on Build GRCh38
rs769602489 Chr.19: 58509909-58509909 on Build GRCh38
rs556623064 Chr.19: 58509824-58509824 on Build GRCh38
rs760053212 Chr.19: 58510039-58510039 on Build GRCh38
rs768460859 Chr.19: 58509934-58509934 on Build GRCh38
rs775443431 Chr.19: 58509910-58509910 on Build GRCh38
rs749204662 Chr.19: 58510000-58510000 on Build GRCh38
rs147622712 Chr.19: 58509996-58509996 on Build GRCh38
rs768528812 Chr.19: 58510003-58510003 on Build GRCh38
rs202202520 Chr.19: 58510033-58510033 on Build GRCh38
rs377653125 Chr.19: 58509863-58509863 on Build GRCh38
rs748319861 Chr.19: 58509892-58509892 on Build GRCh38
rs781164987 Chr.19: 58509999-58509999 on Build GRCh38
rs765863347 Chr.19: 58510034-58510034 on Build GRCh38
rs200114980 Chr.19: 58510011-58510011 on Build GRCh38
rs749190749 Chr.19: 58509915-58509915 on Build GRCh38
rs761535986 Chr.19: 58509957-58509957 on Build GRCh38
rs146682651 Chr.19: 58509970-58509970 on Build GRCh38
rs761272712 Chr.19: 58510010-58510010 on Build GRCh38
rs778025005 Chr.19: 58509981-58509981 on Build GRCh38
rs751159512 Chr.19: 58509825-58509825 on Build GRCh38
rs755301099 Chr.19: 58509877-58509877 on Build GRCh38
rs199844677 Chr.19: 58509874-58509874 on Build GRCh38
rs762861669 Chr.19: 58509822-58509822 on Build GRCh38
rs1003407858 Chr.19: 58509889-58509889 on Build GRCh38
rs770524004 Chr.19: 58509848-58509848 on Build GRCh38
rs757298753 Chr.19: 58509998-58509998 on Build GRCh38
rs371102258 Chr.19: 58509972-58509972 on Build GRCh38
rs747156086 Chr.19: 58509908-58509908 on Build GRCh38
rs113590100 Chr.19: 58509971-58509971 on Build GRCh38
rs143272701 Chr.19: 58509967-58509967 on Build GRCh38
rs777795378 Chr.19: 58509906-58509906 on Build GRCh38
rs754141923 Chr.19: 58509875-58509875 on Build GRCh38
rs771838912 Chr.19: 58510031-58510031 on Build GRCh38
rs774313859 Chr.19: 58509939-58509939 on Build GRCh38
rs750002836 Chr.19: 58509862-58509862 on Build GRCh38

View Full Product Details