Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5947765-5948026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR56A3
Entrez Gene ID
390083
Gene Name
olfactory receptor family 56 subfamily A member 3
Gene Aliases
OR56A2P, OR56A3P, OR56A6
UniGene
Hs.553741
Gene Chromosome Location
Chr.11: 5947347-5948294 on Build GRCh38
Transcripts
NM_001003443.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574518 c.496C>A p.L166I Chr.11: 5947842-5947842 on Build GRCh38
269794 c.551C>T p.A184V Chr.11: 5947897-5947897 on Build GRCh38
376565 c.679C>A p.R227R Chr.11: 5948025-5948025 on Build GRCh38
7171919 c.477G>A p.M159I Chr.11: 5947823-5947823 on Build GRCh38
6488433 c.662C>A p.S221Y Chr.11: 5948008-5948008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773988755 Chr.11: 5947850-5947850 on Build GRCh38
rs768096694 Chr.11: 5947791-5947791 on Build GRCh38
rs748654614 Chr.11: 5947846-5947846 on Build GRCh38
rs559463372 Chr.11: 5947912-5947912 on Build GRCh38
rs946414774 Chr.11: 5947941-5947941 on Build GRCh38
rs760268797 Chr.11: 5947858-5947858 on Build GRCh38
rs986989096 Chr.11: 5947856-5947856 on Build GRCh38
rs749847687 Chr.11: 5947956-5947956 on Build GRCh38
rs374842307 Chr.11: 5947857-5947857 on Build GRCh38
rs780183561 Chr.11: 5947832-5947832 on Build GRCh38
rs758432770 Chr.11: 5947893-5947893 on Build GRCh38
rs781614186 Chr.11: 5947926-5947926 on Build GRCh38
rs765047625 Chr.11: 5947998-5947998 on Build GRCh38
rs755191763 Chr.11: 5947925-5947925 on Build GRCh38
rs774912300 Chr.11: 5947969-5947969 on Build GRCh38
rs778934688 Chr.11: 5947796-5947796 on Build GRCh38
rs865937430 Chr.11: 5948000-5948000 on Build GRCh38
rs746643175 Chr.11: 5947979-5947979 on Build GRCh38
rs749189750 Chr.11: 5947829-5947829 on Build GRCh38
rs756563273 Chr.11: 5947937-5947937 on Build GRCh38
rs751779308 Chr.11: 5947919-5947919 on Build GRCh38
rs201306705 Chr.11: 5947843-5947843 on Build GRCh38
rs373078520 Chr.11: 5947853-5947853 on Build GRCh38
rs761528471 Chr.11: 5947996-5947996 on Build GRCh38
rs761724525 Chr.11: 5947889-5947889 on Build GRCh38
rs761113102 Chr.11: 5947904-5947904 on Build GRCh38
rs776356953 Chr.11: 5947986-5947986 on Build GRCh38
rs532476312 Chr.11: 5947866-5947866 on Build GRCh38
rs750557836 Chr.11: 5947808-5947808 on Build GRCh38
rs763928740 Chr.11: 5947875-5947875 on Build GRCh38
rs955583733 Chr.11: 5947851-5947851 on Build GRCh38
rs768262757 Chr.11: 5947981-5947981 on Build GRCh38
rs747222635 Chr.11: 5947841-5947841 on Build GRCh38
rs10769378 Chr.11: 5947955-5947955 on Build GRCh38
rs769012601 Chr.11: 5947842-5947842 on Build GRCh38
rs540988763 Chr.11: 5947879-5947879 on Build GRCh38
rs771659191 Chr.11: 5947959-5947959 on Build GRCh38
rs750930279 Chr.11: 5947861-5947862 on Build GRCh38
rs766906053 Chr.11: 5947992-5947997 on Build GRCh38
rs370134619 Chr.11: 5947891-5947891 on Build GRCh38
rs376082899 Chr.11: 5947794-5947794 on Build GRCh38
rs780015932 Chr.11: 5947905-5947905 on Build GRCh38
rs770492176 Chr.11: 5947848-5947848 on Build GRCh38
rs748584174 Chr.11: 5947927-5947927 on Build GRCh38
rs201417208 Chr.11: 5947818-5947818 on Build GRCh38

View Full Product Details