Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75232806-75233077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA3
Entrez Gene ID
23163
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Gene Aliases
-
UniGene
Hs.87726
Gene Chromosome Location
Chr.17: 75225464-75262393 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
UniGene
Hs.362817
Gene Chromosome Location
Chr.17: 75205450-75235759 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6082074 c.1018A>G p.R340G Chr.17: 75233065-75233065 on Build GRCh38
3522066 c.1481A>G p.K494R Chr.17: 75232935-75232935 on Build GRCh38
6878132 c.993C>T p.A331A Chr.17: 75232951-75232951 on Build GRCh38
3522067 c.977A>G p.K326R Chr.17: 75232935-75232935 on Build GRCh38
7380752 c.916G>A p.A306T Chr.17: 75232874-75232874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750992371 Chr.17: 75232943-75232943 on Build GRCh38
rs765388958 Chr.17: 75233000-75233000 on Build GRCh38
rs142787021 Chr.17: 75232899-75232899 on Build GRCh38
rs982808612 Chr.17: 75232868-75232868 on Build GRCh38
rs769609522 Chr.17: 75232911-75232911 on Build GRCh38
rs753034446 Chr.17: 75232878-75232878 on Build GRCh38
rs747919003 Chr.17: 75232906-75232906 on Build GRCh38
rs370352066 Chr.17: 75233050-75233050 on Build GRCh38
rs757607656 Chr.17: 75232940-75232940 on Build GRCh38
rs150650595 Chr.17: 75232954-75232954 on Build GRCh38
rs752040574 Chr.17: 75233024-75233024 on Build GRCh38
rs777993444 Chr.17: 75232888-75232888 on Build GRCh38
rs941575230 Chr.17: 75233037-75233037 on Build GRCh38
rs975506262 Chr.17: 75232835-75232835 on Build GRCh38
rs747814899 Chr.17: 75233046-75233046 on Build GRCh38
rs767762494 Chr.17: 75232918-75232918 on Build GRCh38
rs368889675 Chr.17: 75232892-75232892 on Build GRCh38
rs530182331 Chr.17: 75232983-75232983 on Build GRCh38
rs776506979 Chr.17: 75232887-75232887 on Build GRCh38
rs545737851 Chr.17: 75232856-75232856 on Build GRCh38
rs771768808 Chr.17: 75232830-75232834 on Build GRCh38
rs376944043 Chr.17: 75232977-75232977 on Build GRCh38
rs375845560 Chr.17: 75232874-75232874 on Build GRCh38
rs780750356 Chr.17: 75233035-75233035 on Build GRCh38
rs754487507 Chr.17: 75233027-75233027 on Build GRCh38
rs372532025 Chr.17: 75232853-75232853 on Build GRCh38
rs746659980 Chr.17: 75232844-75232844 on Build GRCh38
rs201018542 Chr.17: 75232957-75232957 on Build GRCh38
rs762980962 Chr.17: 75232917-75232917 on Build GRCh38
rs369873595 Chr.17: 75232978-75232978 on Build GRCh38
rs779305795 Chr.17: 75232891-75232891 on Build GRCh38
rs774391267 Chr.17: 75232972-75232972 on Build GRCh38
rs779615147 Chr.17: 75232840-75232840 on Build GRCh38
rs148520579 Chr.17: 75232889-75232889 on Build GRCh38
rs563333903 Chr.17: 75232958-75232958 on Build GRCh38
rs572661092 Chr.17: 75232885-75232885 on Build GRCh38
rs375746742 Chr.17: 75233006-75233006 on Build GRCh38
rs759741322 Chr.17: 75232864-75232864 on Build GRCh38
rs755802307 Chr.17: 75233047-75233047 on Build GRCh38
rs776306989 Chr.17: 75232854-75232854 on Build GRCh38
rs567460945 Chr.17: 75233002-75233002 on Build GRCh38
rs907662063 Chr.17: 75233019-75233019 on Build GRCh38
rs746467614 Chr.17: 75232898-75232898 on Build GRCh38
rs760740948 Chr.17: 75232928-75232928 on Build GRCh38
rs757597966 Chr.17: 75232890-75232890 on Build GRCh38
rs574988208 Chr.17: 75232939-75232939 on Build GRCh38
rs200361674 Chr.17: 75232953-75232953 on Build GRCh38
rs908543464 Chr.17: 75233025-75233025 on Build GRCh38
rs780475013 Chr.17: 75232949-75232949 on Build GRCh38
rs541230324 Chr.17: 75232904-75232904 on Build GRCh38
rs775170459 Chr.17: 75232831-75232831 on Build GRCh38
rs369534023 Chr.17: 75232944-75232944 on Build GRCh38
rs955864300 Chr.17: 75232931-75232931 on Build GRCh38
rs545929051 Chr.17: 75232929-75232929 on Build GRCh38
rs766897830 Chr.17: 75233007-75233007 on Build GRCh38
rs200313531 Chr.17: 75232942-75232942 on Build GRCh38
rs528584321 Chr.17: 75232992-75232992 on Build GRCh38
rs773324515 Chr.17: 75232912-75232912 on Build GRCh38
rs775724512 Chr.17: 75232923-75232923 on Build GRCh38
rs750326333 Chr.17: 75232863-75232863 on Build GRCh38
rs57026156 Chr.17: 75232997-75232997 on Build GRCh38
rs760287609 Chr.17: 75232844-75232844 on Build GRCh38
rs371471946 Chr.17: 75232896-75232896 on Build GRCh38
rs532419654 Chr.17: 75232965-75232965 on Build GRCh38
rs770996694 Chr.17: 75232971-75232971 on Build GRCh38
rs1004425648 Chr.17: 75232830-75232830 on Build GRCh38
rs368637573 Chr.17: 75232893-75232893 on Build GRCh38
rs932994120 Chr.17: 75232991-75232991 on Build GRCh38
rs941769803 Chr.17: 75233030-75233030 on Build GRCh38

View Full Product Details