Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116858094-116858349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIK3
Entrez Gene ID
23387
Gene Name
SIK family kinase 3
Gene Aliases
L19, QSK, SIK-3
UniGene
Hs.167451
Gene Chromosome Location
Chr.11: 116843402-117098453 on Build GRCh38
Transcripts
NM_001281748.1 NM_001281749.1 NM_025164.4 XM_005271481.2 XM_005271482.4 XM_005271484.3 XM_005271485.3 XM_011542721.2 XM_011542722.1 XM_011542723.2 XM_011542724.2 XM_011542725.2 XM_017017424.1 XM_017017425.1 XM_017017426.1 XM_017017427.1

SNP Detail

SNP ID Chromosome Location
rs750633464 Chr.11: 116858257-116858257 on Build GRCh38
rs149318611 Chr.11: 116858268-116858268 on Build GRCh38
rs752642681 Chr.11: 116858315-116858329 on Build GRCh38
rs770511850 Chr.11: 116858222-116858222 on Build GRCh38
rs752525063 Chr.11: 116858260-116858260 on Build GRCh38
rs536178660 Chr.11: 116858283-116858291 on Build GRCh38
rs977767180 Chr.11: 116858182-116858182 on Build GRCh38
rs753895576 Chr.11: 116858153-116858153 on Build GRCh38
rs745572437 Chr.11: 116858235-116858235 on Build GRCh38
rs767454713 Chr.11: 116858314-116858316 on Build GRCh38
rs764166854 Chr.11: 116858146-116858146 on Build GRCh38
rs750685914 Chr.11: 116858179-116858179 on Build GRCh38
rs761357575 Chr.11: 116858298-116858300 on Build GRCh38
rs772521154 Chr.11: 116858294-116858295 on Build GRCh38
rs61738656 Chr.11: 116858197-116858197 on Build GRCh38
rs146353024 Chr.11: 116858256-116858256 on Build GRCh38
rs753241027 Chr.11: 116858133-116858133 on Build GRCh38
rs537893827 Chr.11: 116858295-116858297 on Build GRCh38
rs761276427 Chr.11: 116858134-116858134 on Build GRCh38
rs556463758 Chr.11: 116858199-116858199 on Build GRCh38
rs764722944 Chr.11: 116858297-116858298 on Build GRCh38
rs536284019 Chr.11: 116858240-116858240 on Build GRCh38
rs913467008 Chr.11: 116858170-116858170 on Build GRCh38
rs140534291 Chr.11: 116858162-116858162 on Build GRCh38
rs934700203 Chr.11: 116858314-116858325 on Build GRCh38
rs757452260 Chr.11: 116858159-116858159 on Build GRCh38
rs373958184 Chr.11: 116858241-116858241 on Build GRCh38
rs781024898 Chr.11: 116858181-116858181 on Build GRCh38
rs886373587 Chr.11: 116858282-116858283 on Build GRCh38
rs930482155 Chr.11: 116858269-116858269 on Build GRCh38
rs769106443 Chr.11: 116858310-116858310 on Build GRCh38
rs56247967 Chr.11: 116858173-116858173 on Build GRCh38
rs777001109 Chr.11: 116858312-116858312 on Build GRCh38
rs756706645 Chr.11: 116858272-116858272 on Build GRCh38
rs747314377 Chr.11: 116858304-116858304 on Build GRCh38
rs774632940 Chr.11: 116858288-116858289 on Build GRCh38
rs771881772 Chr.11: 116858291-116858291 on Build GRCh38
rs745446213 Chr.11: 116858284-116858284 on Build GRCh38
rs758636240 Chr.11: 116858258-116858258 on Build GRCh38
rs775912099 Chr.11: 116858296-116858297 on Build GRCh38
rs751346331 Chr.11: 116858303-116858304 on Build GRCh38
rs547832673 Chr.11: 116858129-116858129 on Build GRCh38
rs759945448 Chr.11: 116858125-116858125 on Build GRCh38
rs570258388 Chr.11: 116858238-116858238 on Build GRCh38
rs56161061 Chr.11: 116858126-116858126 on Build GRCh38
rs142486692 Chr.11: 116858229-116858229 on Build GRCh38
rs368405357 Chr.11: 116858264-116858264 on Build GRCh38
rs559308304 Chr.11: 116858250-116858250 on Build GRCh38
rs139073798 Chr.11: 116858259-116858259 on Build GRCh38
rs539858 Chr.11: 116858297-116858297 on Build GRCh38
rs901812167 Chr.11: 116858313-116858313 on Build GRCh38
rs769627045 Chr.11: 116858196-116858196 on Build GRCh38
rs776767431 Chr.11: 116858274-116858274 on Build GRCh38
rs748025109 Chr.11: 116858191-116858191 on Build GRCh38
rs754765058 Chr.11: 116858180-116858180 on Build GRCh38
rs766157160 Chr.11: 116858300-116858301 on Build GRCh38
rs772505893 Chr.11: 116858237-116858237 on Build GRCh38
rs775947586 Chr.11: 116858301-116858301 on Build GRCh38
rs755982568 Chr.11: 116858261-116858261 on Build GRCh38
rs754866280 Chr.11: 116858310-116858311 on Build GRCh38

View Full Product Details