Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120334719-120334974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC58
Entrez Gene ID
116064
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
UniGene
Hs.518084
Gene Chromosome Location
Chr.3: 120324729-120349339 on Build GRCh38
Transcripts
NM_001099678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1418082 c.817C>T p.R273* Chr.3: 120334952-120334952 on Build GRCh38
1036853 c.818G>A p.R273Q Chr.3: 120334951-120334951 on Build GRCh38
1327945 c.861T>G p.L287L Chr.3: 120334908-120334908 on Build GRCh38
3120336 c.871G>A p.G291S Chr.3: 120334898-120334898 on Build GRCh38
6347141 c.806C>A p.T269N Chr.3: 120334963-120334963 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754246322 Chr.3: 120334908-120334908 on Build GRCh38
rs769672555 Chr.3: 120334832-120334832 on Build GRCh38
rs758267452 Chr.3: 120334908-120334910 on Build GRCh38
rs372781365 Chr.3: 120334907-120334907 on Build GRCh38
rs555374371 Chr.3: 120334820-120334820 on Build GRCh38
rs756674201 Chr.3: 120334952-120334952 on Build GRCh38
rs760114843 Chr.3: 120334930-120334930 on Build GRCh38
rs751239277 Chr.3: 120334951-120334951 on Build GRCh38
rs766489976 Chr.3: 120334905-120334905 on Build GRCh38
rs558418628 Chr.3: 120334765-120334765 on Build GRCh38
rs376682808 Chr.3: 120334936-120334936 on Build GRCh38
rs899442886 Chr.3: 120334910-120334910 on Build GRCh38
rs759551906 Chr.3: 120334902-120334902 on Build GRCh38
rs756074886 Chr.3: 120334763-120334763 on Build GRCh38
rs1054778728 Chr.3: 120334803-120334803 on Build GRCh38
rs1005237722 Chr.3: 120334817-120334817 on Build GRCh38
rs765567615 Chr.3: 120334935-120334935 on Build GRCh38
rs894774309 Chr.3: 120334814-120334814 on Build GRCh38
rs34873834 Chr.3: 120334872-120334873 on Build GRCh38
rs777656340 Chr.3: 120334946-120334946 on Build GRCh38
rs773335604 Chr.3: 120334898-120334898 on Build GRCh38
rs535231874 Chr.3: 120334790-120334790 on Build GRCh38
rs762104274 Chr.3: 120334854-120334854 on Build GRCh38
rs368518497 Chr.3: 120334889-120334889 on Build GRCh38
rs372148684 Chr.3: 120334939-120334939 on Build GRCh38
rs367647477 Chr.3: 120334937-120334937 on Build GRCh38
rs566009323 Chr.3: 120334872-120334872 on Build GRCh38
rs1016900829 Chr.3: 120334903-120334903 on Build GRCh38
rs745841879 Chr.3: 120334825-120334825 on Build GRCh38
rs780935763 Chr.3: 120334824-120334824 on Build GRCh38
rs775465114 Chr.3: 120334853-120334853 on Build GRCh38

View Full Product Details