Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149632524-149632797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNA1
Entrez Gene ID
11104
Gene Name
katanin catalytic subunit A1
Gene Aliases
-
UniGene
Hs.450175
Gene Chromosome Location
Chr.6: 149594873-149649018 on Build GRCh38
Transcripts
NM_001204076.1 NM_007044.3 XM_005266804.2 XM_005266805.3 XM_017010207.1 XM_017010208.1 XM_017010209.1 XM_017010210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077601 c.282G>C p.E94D Chr.6: 149632797-149632797 on Build GRCh38
5781282 c.294G>C p.W98C Chr.6: 149632785-149632785 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922185998 Chr.6: 149632551-149632551 on Build GRCh38
rs780582448 Chr.6: 149632574-149632574 on Build GRCh38
rs777638906 Chr.6: 149632735-149632735 on Build GRCh38
rs771062124 Chr.6: 149632754-149632754 on Build GRCh38
rs375351244 Chr.6: 149632726-149632726 on Build GRCh38
rs569603753 Chr.6: 149632603-149632603 on Build GRCh38
rs187986189 Chr.6: 149632756-149632756 on Build GRCh38
rs767403699 Chr.6: 149632659-149632659 on Build GRCh38
rs879204166 Chr.6: 149632683-149632683 on Build GRCh38
rs770023141 Chr.6: 149632769-149632769 on Build GRCh38
rs370751901 Chr.6: 149632712-149632712 on Build GRCh38
rs951211059 Chr.6: 149632734-149632734 on Build GRCh38
rs377538900 Chr.6: 149632708-149632708 on Build GRCh38
rs966615551 Chr.6: 149632622-149632622 on Build GRCh38
rs182517563 Chr.6: 149632641-149632641 on Build GRCh38
rs753412658 Chr.6: 149632709-149632709 on Build GRCh38
rs752426675 Chr.6: 149632718-149632718 on Build GRCh38
rs1054591575 Chr.6: 149632580-149632580 on Build GRCh38
rs958615502 Chr.6: 149632685-149632685 on Build GRCh38
rs533659192 Chr.6: 149632763-149632763 on Build GRCh38
rs1002856639 Chr.6: 149632640-149632640 on Build GRCh38
rs549711307 Chr.6: 149632583-149632583 on Build GRCh38
rs907224763 Chr.6: 149632606-149632606 on Build GRCh38
rs776920052 Chr.6: 149632720-149632720 on Build GRCh38
rs893247003 Chr.6: 149632586-149632586 on Build GRCh38
rs746879529 Chr.6: 149632746-149632746 on Build GRCh38
rs778471145 Chr.6: 149632715-149632715 on Build GRCh38
rs1010362976 Chr.6: 149632596-149632596 on Build GRCh38
rs9322202 Chr.6: 149632699-149632699 on Build GRCh38
rs745997781 Chr.6: 149632762-149632762 on Build GRCh38
rs375960214 Chr.6: 149632775-149632775 on Build GRCh38

View Full Product Details