Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105482193-105482442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUS7
Entrez Gene ID
54517
Gene Name
pseudouridylate synthase 7 (putative)
Gene Aliases
-
UniGene
Hs.520619
Gene Chromosome Location
Chr.7: 105456501-105522270 on Build GRCh38
Transcripts
NM_001318163.1 NM_001318164.1 NM_019042.4 XM_005250462.3 XM_011516336.2 XM_017012367.1 XM_017012368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763364 c.931C>A p.Q311K Chr.7: 105482430-105482430 on Build GRCh38
5764283 c.921-1G>A p.? Chr.7: 105482441-105482441 on Build GRCh38
6307327 c.1015G>A p.E339K Chr.7: 105482346-105482346 on Build GRCh38
5764882 c.935G>A p.R312K Chr.7: 105482426-105482426 on Build GRCh38
1643199 c.998delA p.N333fs*3 Chr.7: 105482363-105482363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771191481 Chr.7: 105482244-105482244 on Build GRCh38
rs367678815 Chr.7: 105482377-105482377 on Build GRCh38
rs779676454 Chr.7: 105482302-105482302 on Build GRCh38
rs1001102159 Chr.7: 105482246-105482246 on Build GRCh38
rs545451019 Chr.7: 105482240-105482240 on Build GRCh38
rs780683591 Chr.7: 105482363-105482363 on Build GRCh38
rs528905477 Chr.7: 105482372-105482372 on Build GRCh38
rs760959784 Chr.7: 105482406-105482406 on Build GRCh38
rs562100610 Chr.7: 105482419-105482419 on Build GRCh38
rs964110373 Chr.7: 105482356-105482356 on Build GRCh38
rs1008395236 Chr.7: 105482286-105482286 on Build GRCh38
rs372333814 Chr.7: 105482407-105482407 on Build GRCh38
rs374833148 Chr.7: 105482282-105482282 on Build GRCh38
rs904201969 Chr.7: 105482245-105482245 on Build GRCh38
rs758254215 Chr.7: 105482291-105482291 on Build GRCh38
rs1019815807 Chr.7: 105482298-105482298 on Build GRCh38
rs775664833 Chr.7: 105482405-105482405 on Build GRCh38
rs192011853 Chr.7: 105482285-105482285 on Build GRCh38
rs757412484 Chr.7: 105482277-105482277 on Build GRCh38
rs948497209 Chr.7: 105482220-105482220 on Build GRCh38
rs764252726 Chr.7: 105482268-105482268 on Build GRCh38
rs200636298 Chr.7: 105482272-105482272 on Build GRCh38
rs558909131 Chr.7: 105482247-105482247 on Build GRCh38
rs931638651 Chr.7: 105482392-105482392 on Build GRCh38
rs561239827 Chr.7: 105482367-105482367 on Build GRCh38
rs746696488 Chr.7: 105482349-105482349 on Build GRCh38
rs747933408 Chr.7: 105482217-105482217 on Build GRCh38
rs374839280 Chr.7: 105482371-105482371 on Build GRCh38
rs1026962602 Chr.7: 105482386-105482386 on Build GRCh38
rs768136334 Chr.7: 105482353-105482353 on Build GRCh38
rs542221978 Chr.7: 105482403-105482403 on Build GRCh38

View Full Product Details