Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40667534-40667807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUMBL
Entrez Gene ID
9253
Gene Name
NUMB like, endocytic adaptor protein
Gene Aliases
CAG3A, CTG3a, NBL, NUMB-R, NUMBLIKE, NUMBR, TNRC23
UniGene
Hs.326953
Gene Chromosome Location
Chr.19: 40665905-40690658 on Build GRCh38
Transcripts
NM_001289979.1 NM_001289980.1 NM_004756.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5657500 c.1710A>C p.P570P Chr.19: 40667588-40667588 on Build GRCh38
5707029 c.1705G>T p.A569S Chr.19: 40667593-40667593 on Build GRCh38
94879 c.1633A>G p.I545V Chr.19: 40667665-40667665 on Build GRCh38
4960164 c.1630G>A p.A544T Chr.19: 40667668-40667668 on Build GRCh38
4654801 c.1708C>T p.P570S Chr.19: 40667590-40667590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756248399 Chr.19: 40667654-40667654 on Build GRCh38
rs74674304 Chr.19: 40667679-40667679 on Build GRCh38
rs1039446474 Chr.19: 40667670-40667670 on Build GRCh38
rs751474840 Chr.19: 40667637-40667637 on Build GRCh38
rs774241843 Chr.19: 40667715-40667715 on Build GRCh38
rs772850434 Chr.19: 40667631-40667631 on Build GRCh38
rs576071736 Chr.19: 40667711-40667711 on Build GRCh38
rs757456532 Chr.19: 40667779-40667779 on Build GRCh38
rs767326374 Chr.19: 40667643-40667643 on Build GRCh38
rs778206793 Chr.19: 40667656-40667656 on Build GRCh38
rs772868730 Chr.19: 40667700-40667700 on Build GRCh38
rs868270667 Chr.19: 40667662-40667662 on Build GRCh38
rs772672013 Chr.19: 40667677-40667677 on Build GRCh38
rs767550517 Chr.19: 40667728-40667728 on Build GRCh38
rs769498350 Chr.19: 40667625-40667625 on Build GRCh38
rs752873457 Chr.19: 40667647-40667647 on Build GRCh38
rs769229011 Chr.19: 40667694-40667694 on Build GRCh38
rs776165005 Chr.19: 40667616-40667616 on Build GRCh38
rs746388730 Chr.19: 40667676-40667676 on Build GRCh38
rs754134316 Chr.19: 40667774-40667774 on Build GRCh38
rs113076086 Chr.19: 40667624-40667624 on Build GRCh38
rs747726463 Chr.19: 40667685-40667685 on Build GRCh38
rs572713701 Chr.19: 40667612-40667612 on Build GRCh38
rs73546875 Chr.19: 40667743-40667743 on Build GRCh38
rs754046549 Chr.19: 40667661-40667661 on Build GRCh38
rs374888940 Chr.19: 40667615-40667615 on Build GRCh38
rs554497777 Chr.19: 40667611-40667611 on Build GRCh38
rs900605844 Chr.19: 40667669-40667670 on Build GRCh38
rs752675146 Chr.19: 40667729-40667729 on Build GRCh38
rs779291267 Chr.19: 40667598-40667598 on Build GRCh38
rs369374254 Chr.19: 40667744-40667744 on Build GRCh38
rs974650272 Chr.19: 40667686-40667686 on Build GRCh38
rs142060332 Chr.19: 40667553-40667553 on Build GRCh38
rs543463869 Chr.19: 40667736-40667736 on Build GRCh38
rs762744056 Chr.19: 40667634-40667634 on Build GRCh38
rs566641405 Chr.19: 40667788-40667788 on Build GRCh38
rs372917525 Chr.19: 40667756-40667756 on Build GRCh38
rs1026313838 Chr.19: 40667786-40667786 on Build GRCh38
rs963484272 Chr.19: 40667777-40667777 on Build GRCh38
rs762547083 Chr.19: 40667706-40667706 on Build GRCh38
rs199881260 Chr.19: 40667780-40667780 on Build GRCh38
rs555115582 Chr.19: 40667638-40667638 on Build GRCh38
rs757574949 Chr.19: 40667669-40667669 on Build GRCh38
rs747088284 Chr.19: 40667586-40667591 on Build GRCh38
rs1010494062 Chr.19: 40667629-40667629 on Build GRCh38
rs759232395 Chr.19: 40667723-40667723 on Build GRCh38
rs747537068 Chr.19: 40667597-40667597 on Build GRCh38
rs779284323 Chr.19: 40667672-40667672 on Build GRCh38
rs766143681 Chr.19: 40667636-40667636 on Build GRCh38
rs771417907 Chr.19: 40667591-40667591 on Build GRCh38
rs202221260 Chr.19: 40667787-40667787 on Build GRCh38

View Full Product Details