Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41729592-41729859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
UniGene
Hs.116419
Gene Chromosome Location
Chr.22: 41699503-41799456 on Build GRCh38
Transcripts
NM_152513.3 XM_011529935.1 XM_011529936.1 XM_011529937.1 XM_011529938.1 XM_011529939.1 XM_011529940.1 XM_011529941.1 XM_011529942.2 XM_011529943.1 XM_011529944.1 XM_011529945.2 XM_011529946.1 XM_011529947.1 XM_011529948.2 XM_011529949.1 XM_011529950.1 XM_011529952.1 XM_011529955.1 XM_011529956.2 XM_017028633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3153637 c.950C>T p.A317V Chr.22: 41729750-41729750 on Build GRCh38
4104506 c.128A>G p.H43R Chr.22: 41729708-41729708 on Build GRCh38
5499445 c.171G>A p.A57A Chr.22: 41729751-41729751 on Build GRCh38
387294 c.967G>A p.A323T Chr.22: 41729767-41729767 on Build GRCh38
4300154 c.170C>T p.A57V Chr.22: 41729750-41729750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769346948 Chr.22: 41729811-41729811 on Build GRCh38
rs1001788351 Chr.22: 41729830-41729830 on Build GRCh38
rs753698490 Chr.22: 41729792-41729792 on Build GRCh38
rs769115806 Chr.22: 41729716-41729716 on Build GRCh38
rs762737705 Chr.22: 41729621-41729621 on Build GRCh38
rs566045675 Chr.22: 41729766-41729766 on Build GRCh38
rs749489352 Chr.22: 41729659-41729659 on Build GRCh38
rs777584660 Chr.22: 41729657-41729657 on Build GRCh38
rs745882325 Chr.22: 41729800-41729800 on Build GRCh38
rs778767400 Chr.22: 41729817-41729817 on Build GRCh38
rs752598918 Chr.22: 41729783-41729783 on Build GRCh38
rs755926756 Chr.22: 41729655-41729655 on Build GRCh38
rs767309925 Chr.22: 41729777-41729777 on Build GRCh38
rs757556842 Chr.22: 41729661-41729661 on Build GRCh38
rs201481793 Chr.22: 41729638-41729638 on Build GRCh38
rs975144462 Chr.22: 41729816-41729816 on Build GRCh38
rs778853271 Chr.22: 41729669-41729669 on Build GRCh38
rs539486418 Chr.22: 41729809-41729809 on Build GRCh38
rs376715038 Chr.22: 41729775-41729775 on Build GRCh38
rs770536781 Chr.22: 41729754-41729754 on Build GRCh38
rs758416363 Chr.22: 41729799-41729801 on Build GRCh38
rs781196206 Chr.22: 41729630-41729630 on Build GRCh38
rs758532774 Chr.22: 41729801-41729801 on Build GRCh38
rs370424156 Chr.22: 41729625-41729625 on Build GRCh38
rs1019286277 Chr.22: 41729789-41729789 on Build GRCh38
rs765442355 Chr.22: 41729668-41729670 on Build GRCh38
rs757351698 Chr.22: 41729794-41729794 on Build GRCh38
rs779206647 Chr.22: 41729798-41729798 on Build GRCh38
rs904909987 Chr.22: 41729814-41729814 on Build GRCh38
rs529584550 Chr.22: 41729680-41729680 on Build GRCh38
rs748552073 Chr.22: 41729827-41729827 on Build GRCh38
rs750135763 Chr.22: 41729619-41729619 on Build GRCh38
rs766123638 Chr.22: 41729751-41729751 on Build GRCh38
rs763862544 Chr.22: 41729791-41729791 on Build GRCh38
rs780324336 Chr.22: 41729807-41729807 on Build GRCh38
rs1026288941 Chr.22: 41729721-41729721 on Build GRCh38
rs776083441 Chr.22: 41729713-41729713 on Build GRCh38
rs958086464 Chr.22: 41729829-41729829 on Build GRCh38
rs772397733 Chr.22: 41729702-41729702 on Build GRCh38
rs777187865 Chr.22: 41729742-41729742 on Build GRCh38
rs199660076 Chr.22: 41729750-41729750 on Build GRCh38
rs747333551 Chr.22: 41729714-41729714 on Build GRCh38
rs759100618 Chr.22: 41729767-41729767 on Build GRCh38
rs748770310 Chr.22: 41729624-41729624 on Build GRCh38
rs756092617 Chr.22: 41729785-41729785 on Build GRCh38
rs375386055 Chr.22: 41729799-41729799 on Build GRCh38
rs995791498 Chr.22: 41729685-41729685 on Build GRCh38
rs752778880 Chr.22: 41729777-41729778 on Build GRCh38

View Full Product Details