Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139525806-139526031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507291
Entrez Gene ID
100507291
Gene Name
uncharacterized LOC100507291
Gene Aliases
-
UniGene
Hs.699071
Gene Chromosome Location
Chr.3: 139389803-139583319 on Build GRCh38
Transcripts
NR_121609.1 NM_001130993.1 NM_002899.3

Gene Symbol
RBP1
Entrez Gene ID
5947
Gene Name
retinol binding protein 1
Gene Aliases
CRABP-I, CRBP, CRBP1, CRBPI, RBPC
UniGene
Hs.529571
Gene Chromosome Location
Chr.3: 139517434-139539829 on Build GRCh38
Transcripts
NR_121609.1 NM_001130993.1 NM_002899.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6304431 c.440G>T p.S147I Chr.3: 139525901-139525901 on Build GRCh38
445697 c.442G>A p.E148K Chr.3: 139525899-139525899 on Build GRCh38
3124347 c.448G>T p.G150C Chr.3: 139525893-139525893 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs566181717 Chr.3: 139525915-139525915 on Build GRCh38
rs979390367 Chr.3: 139525984-139525984 on Build GRCh38
rs755923710 Chr.3: 139525825-139525825 on Build GRCh38
rs373816398 Chr.3: 139525844-139525844 on Build GRCh38
rs781567003 Chr.3: 139525951-139525951 on Build GRCh38
rs374499017 Chr.3: 139525888-139525888 on Build GRCh38
rs1044133575 Chr.3: 139525833-139525833 on Build GRCh38
rs778211792 Chr.3: 139525832-139525832 on Build GRCh38
rs188512844 Chr.3: 139525864-139525864 on Build GRCh38
rs768322869 Chr.3: 139525884-139525884 on Build GRCh38
rs582786 Chr.3: 139525916-139525916 on Build GRCh38
rs200344887 Chr.3: 139525901-139525901 on Build GRCh38
rs370198983 Chr.3: 139525851-139525851 on Build GRCh38
rs575162633 Chr.3: 139525948-139525948 on Build GRCh38
rs751710707 Chr.3: 139525944-139525944 on Build GRCh38
rs151304588 Chr.3: 139525845-139525845 on Build GRCh38
rs549585 Chr.3: 139525919-139525919 on Build GRCh38
rs77537608 Chr.3: 139525876-139525876 on Build GRCh38
rs554905154 Chr.3: 139525930-139525930 on Build GRCh38
rs1000962308 Chr.3: 139525982-139525982 on Build GRCh38
rs910639497 Chr.3: 139525986-139525986 on Build GRCh38
rs553535378 Chr.3: 139525851-139525851 on Build GRCh38
rs34689690 Chr.3: 139525967-139525968 on Build GRCh38
rs140569831 Chr.3: 139525977-139525977 on Build GRCh38
rs6808946 Chr.3: 139525920-139525920 on Build GRCh38

View Full Product Details