Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35180581-35180854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0391
Entrez Gene ID
9692
Gene Name
KIAA0391
Gene Aliases
MRPP3, PRORP
UniGene
Hs.458487
Gene Chromosome Location
Chr.14: 35121855-35274078 on Build GRCh38
Transcripts
NM_001256678.1 NM_001256679.1 NM_001256680.1 NM_001256681.1 NM_014672.3 XM_005268237.3 XM_011537409.2 XM_011537410.2 XM_017021835.1 XM_017021836.1 XM_017021837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4695262 c.1174A>C p.K392Q Chr.14: 35180676-35180676 on Build GRCh38
1369669 c.1214A>T p.D405V Chr.14: 35180716-35180716 on Build GRCh38
4695263 c.1262G>A p.R421H Chr.14: 35180764-35180764 on Build GRCh38
6538077 c.1231C>G p.L411V Chr.14: 35180733-35180733 on Build GRCh38
3814736 c.1176G>C p.K392N Chr.14: 35180678-35180678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768432165 Chr.14: 35180816-35180816 on Build GRCh38
rs755178679 Chr.14: 35180673-35180673 on Build GRCh38
rs745344056 Chr.14: 35180806-35180806 on Build GRCh38
rs778635852 Chr.14: 35180799-35180799 on Build GRCh38
rs764714439 Chr.14: 35180695-35180696 on Build GRCh38
rs761804328 Chr.14: 35180827-35180827 on Build GRCh38
rs147065101 Chr.14: 35180763-35180763 on Build GRCh38
rs771904261 Chr.14: 35180807-35180807 on Build GRCh38
rs374891367 Chr.14: 35180812-35180812 on Build GRCh38
rs779708793 Chr.14: 35180809-35180809 on Build GRCh38
rs778289900 Chr.14: 35180692-35180692 on Build GRCh38
rs567593074 Chr.14: 35180706-35180706 on Build GRCh38
rs184890203 Chr.14: 35180831-35180831 on Build GRCh38
rs200805783 Chr.14: 35180755-35180755 on Build GRCh38
rs757828003 Chr.14: 35180798-35180798 on Build GRCh38
rs148259590 Chr.14: 35180737-35180737 on Build GRCh38
rs951768852 Chr.14: 35180628-35180628 on Build GRCh38
rs756575559 Chr.14: 35180774-35180774 on Build GRCh38
rs143102943 Chr.14: 35180743-35180743 on Build GRCh38
rs965942319 Chr.14: 35180728-35180728 on Build GRCh38
rs368620353 Chr.14: 35180652-35180652 on Build GRCh38
rs776592330 Chr.14: 35180738-35180738 on Build GRCh38
rs756627761 Chr.14: 35180691-35180691 on Build GRCh38
rs372115861 Chr.14: 35180762-35180762 on Build GRCh38
rs143121800 Chr.14: 35180694-35180694 on Build GRCh38
rs778144088 Chr.14: 35180768-35180768 on Build GRCh38
rs781602642 Chr.14: 35180675-35180675 on Build GRCh38
rs776449062 Chr.14: 35180824-35180824 on Build GRCh38
rs564548984 Chr.14: 35180613-35180613 on Build GRCh38
rs547075844 Chr.14: 35180704-35180704 on Build GRCh38
rs748513487 Chr.14: 35180685-35180685 on Build GRCh38
rs750909383 Chr.14: 35180819-35180822 on Build GRCh38
rs756457316 Chr.14: 35180764-35180764 on Build GRCh38
rs754311089 Chr.14: 35180784-35180784 on Build GRCh38
rs745576669 Chr.14: 35180703-35180703 on Build GRCh38
rs760500911 Chr.14: 35180724-35180724 on Build GRCh38
rs962085727 Chr.14: 35180817-35180817 on Build GRCh38
rs921785076 Chr.14: 35180753-35180753 on Build GRCh38
rs149659430 Chr.14: 35180618-35180618 on Build GRCh38

View Full Product Details