Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 84149419-84149644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP162
Entrez Gene ID
22832
Gene Name
centrosomal protein 162
Gene Aliases
C6orf84, KIAA1009, QN1
UniGene
Hs.485865
Gene Chromosome Location
Chr.6: 84124241-84227889 on Build GRCh38
Transcripts
NM_001286206.1 NM_014895.3 XM_005248674.3 XM_006715380.2 XM_011535592.2 XM_017010483.1 XM_017010484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623283 c.3742G>T p.E1248* Chr.6: 84149591-84149591 on Build GRCh38
261581 c.3725C>A p.S1242Y Chr.6: 84149608-84149608 on Build GRCh38
5988108 c.3749A>G p.N1250S Chr.6: 84149584-84149584 on Build GRCh38
7429785 c.3689T>G p.I1230R Chr.6: 84149644-84149644 on Build GRCh38
3358058 c.3752G>A p.R1251H Chr.6: 84149581-84149581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761319027 Chr.6: 84149529-84149529 on Build GRCh38
rs374289223 Chr.6: 84149577-84149577 on Build GRCh38
rs540885229 Chr.6: 84149564-84149564 on Build GRCh38
rs756224926 Chr.6: 84149608-84149608 on Build GRCh38
rs1039697043 Chr.6: 84149552-84149552 on Build GRCh38
rs889630664 Chr.6: 84149600-84149600 on Build GRCh38
rs753119513 Chr.6: 84149591-84149591 on Build GRCh38
rs777525586 Chr.6: 84149547-84149548 on Build GRCh38
rs75251993 Chr.6: 84149457-84149457 on Build GRCh38
rs150383742 Chr.6: 84149581-84149581 on Build GRCh38
rs371342692 Chr.6: 84149558-84149558 on Build GRCh38
rs776518627 Chr.6: 84149516-84149516 on Build GRCh38
rs781364195 Chr.6: 84149582-84149582 on Build GRCh38
rs966981094 Chr.6: 84149456-84149456 on Build GRCh38
rs913722447 Chr.6: 84149454-84149454 on Build GRCh38
rs944012443 Chr.6: 84149498-84149498 on Build GRCh38
rs892772548 Chr.6: 84149515-84149515 on Build GRCh38
rs912653002 Chr.6: 84149488-84149488 on Build GRCh38
rs546256904 Chr.6: 84149519-84149519 on Build GRCh38
rs752038590 Chr.6: 84149566-84149566 on Build GRCh38
rs747299684 Chr.6: 84149607-84149608 on Build GRCh38
rs764879222 Chr.6: 84149543-84149543 on Build GRCh38
rs751434157 Chr.6: 84149504-84149504 on Build GRCh38
rs758142351 Chr.6: 84149535-84149535 on Build GRCh38
rs1008097706 Chr.6: 84149602-84149604 on Build GRCh38
rs752523174 Chr.6: 84149528-84149529 on Build GRCh38
rs1004386163 Chr.6: 84149602-84149602 on Build GRCh38
rs147140705 Chr.6: 84149556-84149556 on Build GRCh38
rs1053224276 Chr.6: 84149513-84149513 on Build GRCh38

View Full Product Details