Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102593296-102593569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP20
Entrez Gene ID
9313
Gene Name
matrix metallopeptidase 20
Gene Aliases
AI2A2, MMP-20
UniGene
Hs.591946
Gene Chromosome Location
Chr.11: 102576835-102625332 on Build GRCh38
Transcripts
NM_004771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2108230 c.1219C>T p.L407F Chr.11: 102593467-102593467 on Build GRCh38
4091233 c.1170C>T p.H390H Chr.11: 102593516-102593516 on Build GRCh38
685787 c.1224C>G p.F408L Chr.11: 102593462-102593462 on Build GRCh38
6713840 c.1171G>A p.V391M Chr.11: 102593515-102593515 on Build GRCh38
3868276 c.1218C>T p.T406T Chr.11: 102593468-102593468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551438274 Chr.11: 102593433-102593433 on Build GRCh38
rs747080892 Chr.11: 102593445-102593445 on Build GRCh38
rs770005598 Chr.11: 102593485-102593485 on Build GRCh38
rs537663295 Chr.11: 102593515-102593515 on Build GRCh38
rs1045606470 Chr.11: 102593386-102593386 on Build GRCh38
rs866572582 Chr.11: 102593546-102593547 on Build GRCh38
rs753538852 Chr.11: 102593519-102593519 on Build GRCh38
rs768663486 Chr.11: 102593447-102593447 on Build GRCh38
rs373856753 Chr.11: 102593418-102593418 on Build GRCh38
rs886047545 Chr.11: 102593431-102593431 on Build GRCh38
rs762087055 Chr.11: 102593474-102593474 on Build GRCh38
rs766873878 Chr.11: 102593502-102593502 on Build GRCh38
rs760210732 Chr.11: 102593513-102593513 on Build GRCh38
rs751707514 Chr.11: 102593488-102593488 on Build GRCh38
rs150191942 Chr.11: 102593467-102593467 on Build GRCh38
rs192363990 Chr.11: 102593340-102593340 on Build GRCh38
rs951692136 Chr.11: 102593359-102593359 on Build GRCh38
rs377198461 Chr.11: 102593436-102593436 on Build GRCh38
rs559285776 Chr.11: 102593397-102593397 on Build GRCh38
rs908227750 Chr.11: 102593532-102593532 on Build GRCh38
rs775319636 Chr.11: 102593440-102593440 on Build GRCh38
rs370708788 Chr.11: 102593497-102593497 on Build GRCh38
rs375759479 Chr.11: 102593498-102593498 on Build GRCh38
rs2292732 Chr.11: 102593332-102593332 on Build GRCh38
rs756904716 Chr.11: 102593544-102593544 on Build GRCh38
rs777541124 Chr.11: 102593416-102593416 on Build GRCh38
rs780801800 Chr.11: 102593333-102593333 on Build GRCh38
rs548985085 Chr.11: 102593365-102593365 on Build GRCh38
rs766812216 Chr.11: 102593410-102593410 on Build GRCh38
rs564059592 Chr.11: 102593319-102593319 on Build GRCh38
rs757106372 Chr.11: 102593422-102593422 on Build GRCh38
rs907581668 Chr.11: 102593371-102593371 on Build GRCh38
rs752051781 Chr.11: 102593510-102593510 on Build GRCh38
rs767478911 Chr.11: 102593547-102593547 on Build GRCh38
rs534530637 Chr.11: 102593516-102593516 on Build GRCh38
rs750923051 Chr.11: 102593398-102593398 on Build GRCh38
rs199839391 Chr.11: 102593414-102593414 on Build GRCh38
rs755615025 Chr.11: 102593415-102593415 on Build GRCh38
rs975514141 Chr.11: 102593378-102593378 on Build GRCh38
rs758870448 Chr.11: 102593405-102593405 on Build GRCh38
rs771216203 Chr.11: 102593372-102593372 on Build GRCh38
rs1032788777 Chr.11: 102593320-102593320 on Build GRCh38
rs184655088 Chr.11: 102593548-102593548 on Build GRCh38
rs763010161 Chr.11: 102593364-102593364 on Build GRCh38
rs778920093 Chr.11: 102593430-102593430 on Build GRCh38

View Full Product Details