Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40737579-40737852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPKC
Entrez Gene ID
80271
Gene Name
inositol-trisphosphate 3-kinase C
Gene Aliases
IP3-3KC, IP3KC
UniGene
Hs.515415
Gene Chromosome Location
Chr.19: 40717103-40740860 on Build GRCh38
Transcripts
NM_025194.2 XM_017027324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1136197 c.1814C>G p.A605G Chr.19: 40737735-40737735 on Build GRCh38
3534194 c.1787T>A p.V596E Chr.19: 40737708-40737708 on Build GRCh38
5855844 c.1798G>A p.E600K Chr.19: 40737719-40737719 on Build GRCh38
5389814 c.1842C>T p.T614T Chr.19: 40737763-40737763 on Build GRCh38
4078359 c.1806T>C p.L602L Chr.19: 40737727-40737727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765825251 Chr.19: 40737795-40737795 on Build GRCh38
rs754789349 Chr.19: 40737723-40737723 on Build GRCh38
rs61159620 Chr.19: 40737754-40737754 on Build GRCh38
rs137938894 Chr.19: 40737696-40737696 on Build GRCh38
rs527997855 Chr.19: 40737809-40737809 on Build GRCh38
rs772571483 Chr.19: 40737774-40737774 on Build GRCh38
rs143402835 Chr.19: 40737729-40737729 on Build GRCh38
rs140693572 Chr.19: 40737750-40737750 on Build GRCh38
rs764066908 Chr.19: 40737643-40737643 on Build GRCh38
rs376711814 Chr.19: 40737788-40737788 on Build GRCh38
rs773097589 Chr.19: 40737668-40737668 on Build GRCh38
rs76877448 Chr.19: 40737666-40737666 on Build GRCh38
rs904288189 Chr.19: 40737728-40737728 on Build GRCh38
rs1055346078 Chr.19: 40737674-40737674 on Build GRCh38
rs146426770 Chr.19: 40737613-40737613 on Build GRCh38
rs35178372 Chr.19: 40737619-40737620 on Build GRCh38
rs763299336 Chr.19: 40737670-40737670 on Build GRCh38
rs1006604976 Chr.19: 40737781-40737781 on Build GRCh38
rs781137361 Chr.19: 40737650-40737650 on Build GRCh38
rs759778406 Chr.19: 40737773-40737773 on Build GRCh38
rs201578269 Chr.19: 40737739-40737739 on Build GRCh38
rs771262579 Chr.19: 40737671-40737671 on Build GRCh38
rs774740783 Chr.19: 40737772-40737772 on Build GRCh38
rs749301440 Chr.19: 40737761-40737761 on Build GRCh38
rs759945308 Chr.19: 40737681-40737681 on Build GRCh38
rs747986926 Chr.19: 40737656-40737656 on Build GRCh38
rs775716144 Chr.19: 40737776-40737776 on Build GRCh38
rs775582671 Chr.19: 40737802-40737802 on Build GRCh38
rs751173291 Chr.19: 40737707-40737707 on Build GRCh38
rs749888176 Chr.19: 40737695-40737695 on Build GRCh38
rs774900515 Chr.19: 40737677-40737677 on Build GRCh38
rs761304160 Chr.19: 40737787-40737787 on Build GRCh38
rs779650466 Chr.19: 40737706-40737706 on Build GRCh38
rs371760094 Chr.19: 40737803-40737803 on Build GRCh38
rs544984128 Chr.19: 40737798-40737798 on Build GRCh38
rs754413835 Chr.19: 40737794-40737794 on Build GRCh38
rs762532807 Chr.19: 40737822-40737822 on Build GRCh38
rs960604761 Chr.19: 40737820-40737820 on Build GRCh38
rs369988625 Chr.19: 40737800-40737800 on Build GRCh38
rs998434316 Chr.19: 40737744-40737744 on Build GRCh38
rs371102280 Chr.19: 40737639-40737639 on Build GRCh38
rs374826896 Chr.19: 40737808-40737808 on Build GRCh38
rs576116150 Chr.19: 40737682-40737682 on Build GRCh38
rs1028267170 Chr.19: 40737804-40737804 on Build GRCh38
rs369117388 Chr.19: 40737687-40737687 on Build GRCh38
rs772819126 Chr.19: 40737790-40737790 on Build GRCh38
rs1005403493 Chr.19: 40737823-40737823 on Build GRCh38
rs145921796 Chr.19: 40737766-40737766 on Build GRCh38
rs753273956 Chr.19: 40737685-40737685 on Build GRCh38

View Full Product Details