Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202941798-202942059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARF
Entrez Gene ID
79800
Gene Name
calcium responsive transcription factor
Gene Aliases
ALS2CR8, NYD-SP24
UniGene
Hs.444982
Gene Chromosome Location
Chr.2: 202912218-202987856 on Build GRCh38
Transcripts
NM_001104586.2 NM_001282910.1 NM_001282911.1 NM_001282912.1 NM_001322427.1 NM_001322428.1 NM_001322429.1 NM_001322430.1 NM_024744.15 NR_136330.1 NR_136331.1 XM_005246858.3 XM_005246859.3 XM_006712760.2 XM_011511867.2 XM_011511868.2 XM_017004961.1 XM_017004963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2908155 c.11C>G p.S4C Chr.2: 202941913-202941913 on Build GRCh38
1404617 c.53_54insA p.T20fs*7 Chr.2: 202941955-202941956 on Build GRCh38
5353863 c.54delA p.T20fs*9 Chr.2: 202941956-202941956 on Build GRCh38
3576449 c.70G>A p.V24I Chr.2: 202941972-202941972 on Build GRCh38
362935 c.60C>G p.T20T Chr.2: 202941962-202941962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368948266 Chr.2: 202941897-202941897 on Build GRCh38
rs772965397 Chr.2: 202941831-202941831 on Build GRCh38
rs548204203 Chr.2: 202941997-202941997 on Build GRCh38
rs748429967 Chr.2: 202941933-202941933 on Build GRCh38
rs760706765 Chr.2: 202941944-202941944 on Build GRCh38
rs376838852 Chr.2: 202941905-202941905 on Build GRCh38
rs753097803 Chr.2: 202941974-202941974 on Build GRCh38
rs372946549 Chr.2: 202941983-202941983 on Build GRCh38
rs149943854 Chr.2: 202941943-202941943 on Build GRCh38
rs772264440 Chr.2: 202941937-202941937 on Build GRCh38
rs775732255 Chr.2: 202941930-202941930 on Build GRCh38
rs944996128 Chr.2: 202941910-202941910 on Build GRCh38
rs751191242 Chr.2: 202941864-202941864 on Build GRCh38
rs776427133 Chr.2: 202941925-202941925 on Build GRCh38
rs376249279 Chr.2: 202941973-202941973 on Build GRCh38
rs75869289 Chr.2: 202942001-202942001 on Build GRCh38
rs372267166 Chr.2: 202941899-202941899 on Build GRCh38
rs777140869 Chr.2: 202941950-202941950 on Build GRCh38
rs772545369 Chr.2: 202941927-202941927 on Build GRCh38
rs755597893 Chr.2: 202941987-202941987 on Build GRCh38
rs747105238 Chr.2: 202942025-202942025 on Build GRCh38
rs184892301 Chr.2: 202941890-202941890 on Build GRCh38
rs779997173 Chr.2: 202941903-202941903 on Build GRCh38
rs528810440 Chr.2: 202941848-202941848 on Build GRCh38
rs770978007 Chr.2: 202942026-202942026 on Build GRCh38
rs749980405 Chr.2: 202941984-202941984 on Build GRCh38
rs533756170 Chr.2: 202942017-202942017 on Build GRCh38
rs772315852 Chr.2: 202942014-202942014 on Build GRCh38
rs373506413 Chr.2: 202941971-202941971 on Build GRCh38
rs778821072 Chr.2: 202942005-202942005 on Build GRCh38
rs112546738 Chr.2: 202941845-202941845 on Build GRCh38
rs780136774 Chr.2: 202941950-202941951 on Build GRCh38
rs577882043 Chr.2: 202941936-202941936 on Build GRCh38
rs766817483 Chr.2: 202941945-202941945 on Build GRCh38
rs369170727 Chr.2: 202941978-202941978 on Build GRCh38
rs756963927 Chr.2: 202941866-202941866 on Build GRCh38
rs747910450 Chr.2: 202942012-202942012 on Build GRCh38
rs754861276 Chr.2: 202942004-202942004 on Build GRCh38
rs764619481 Chr.2: 202941953-202941953 on Build GRCh38

View Full Product Details