Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 157592845-157593102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC14
Entrez Gene ID
79683
Gene Name
zinc finger DHHC-type containing 14
Gene Aliases
NEW1CP
UniGene
Hs.596214
Gene Chromosome Location
Chr.6: 157381525-157678114 on Build GRCh38
Transcripts
NM_024630.2 NM_153746.1 XM_005267146.3 XM_011536120.2 XM_011536123.2 XM_017011307.1 XM_017011308.1 XM_017011309.1 XM_017011310.1 XM_017011311.1 XM_017011312.1 XM_017011313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470900 c.426T>G p.S142R Chr.6: 157593007-157593007 on Build GRCh38
6173051 c.440G>C p.R147P Chr.6: 157593021-157593021 on Build GRCh38
6105768 c.430G>T p.G144W Chr.6: 157593011-157593011 on Build GRCh38
7104637 c.440G>A p.R147H Chr.6: 157593021-157593021 on Build GRCh38
4635349 c.497A>G p.Y166C Chr.6: 157593078-157593078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891461646 Chr.6: 157593000-157593000 on Build GRCh38
rs779898396 Chr.6: 157593049-157593049 on Build GRCh38
rs111339992 Chr.6: 157592865-157592865 on Build GRCh38
rs772135347 Chr.6: 157592971-157592973 on Build GRCh38
rs1000350093 Chr.6: 157592941-157592941 on Build GRCh38
rs189112430 Chr.6: 157592992-157592992 on Build GRCh38
rs779702394 Chr.6: 157592956-157592956 on Build GRCh38
rs537665521 Chr.6: 157593021-157593021 on Build GRCh38
rs778832026 Chr.6: 157592964-157592965 on Build GRCh38
rs748780428 Chr.6: 157593064-157593064 on Build GRCh38
rs1057450867 Chr.6: 157592874-157592874 on Build GRCh38
rs941825601 Chr.6: 157592893-157592893 on Build GRCh38
rs867888270 Chr.6: 157592951-157592951 on Build GRCh38
rs778510507 Chr.6: 157593068-157593068 on Build GRCh38
rs917834187 Chr.6: 157592869-157592869 on Build GRCh38
rs773006530 Chr.6: 157592990-157592990 on Build GRCh38
rs759574099 Chr.6: 157592998-157592998 on Build GRCh38
rs755811326 Chr.6: 157593036-157593036 on Build GRCh38
rs200700905 Chr.6: 157593024-157593024 on Build GRCh38
rs548877967 Chr.6: 157592876-157592876 on Build GRCh38
rs201842953 Chr.6: 157592982-157592982 on Build GRCh38
rs534573862 Chr.6: 157592922-157592922 on Build GRCh38
rs747999954 Chr.6: 157592967-157592967 on Build GRCh38
rs181446232 Chr.6: 157592993-157592993 on Build GRCh38
rs199614079 Chr.6: 157592963-157592963 on Build GRCh38
rs748860485 Chr.6: 157592957-157592957 on Build GRCh38
rs753094454 Chr.6: 157593003-157593003 on Build GRCh38
rs971952713 Chr.6: 157593012-157593012 on Build GRCh38
rs1028681801 Chr.6: 157592945-157592945 on Build GRCh38
rs748151450 Chr.6: 157592965-157592965 on Build GRCh38
rs567567123 Chr.6: 157592885-157592885 on Build GRCh38
rs754409889 Chr.6: 157593065-157593065 on Build GRCh38
rs200194288 Chr.6: 157592962-157592962 on Build GRCh38
rs760821344 Chr.6: 157592991-157592991 on Build GRCh38
rs1039213807 Chr.6: 157592924-157592924 on Build GRCh38
rs553001459 Chr.6: 157592872-157592872 on Build GRCh38
rs758955843 Chr.6: 157593011-157593011 on Build GRCh38
rs1024322242 Chr.6: 157592906-157592918 on Build GRCh38
rs909075780 Chr.6: 157592886-157592886 on Build GRCh38

View Full Product Details