Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39600477-39600714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1I
Entrez Gene ID
8911
Gene Name
calcium voltage-gated channel subunit alpha1 I
Gene Aliases
Cav3.3, ca(v)3.3
UniGene
Hs.125116
Gene Chromosome Location
Chr.22: 39570753-39689737 on Build GRCh38
Transcripts
NM_001003406.1 NM_021096.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7254338 c.415delA p.I139fs*45 Chr.22: 39600586-39600586 on Build GRCh38
1593042 c.391G>T p.V131L Chr.22: 39600562-39600562 on Build GRCh38
5603042 c.380C>T p.A127V Chr.22: 39600551-39600551 on Build GRCh38
3842710 c.416T>G p.I139S Chr.22: 39600587-39600587 on Build GRCh38
5603041 c.380C>T p.A127V Chr.22: 39600551-39600551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767779428 Chr.22: 39600585-39600585 on Build GRCh38
rs756481754 Chr.22: 39600625-39600625 on Build GRCh38
rs753045602 Chr.22: 39600618-39600618 on Build GRCh38
rs374150031 Chr.22: 39600666-39600666 on Build GRCh38
rs757987337 Chr.22: 39600649-39600649 on Build GRCh38
rs751780724 Chr.22: 39600564-39600564 on Build GRCh38
rs779653256 Chr.22: 39600655-39600655 on Build GRCh38
rs747940193 Chr.22: 39600663-39600663 on Build GRCh38
rs58395851 Chr.22: 39600553-39600553 on Build GRCh38
rs577831334 Chr.22: 39600546-39600546 on Build GRCh38
rs377381817 Chr.22: 39600507-39600507 on Build GRCh38
rs866406659 Chr.22: 39600574-39600574 on Build GRCh38
rs375201237 Chr.22: 39600512-39600512 on Build GRCh38
rs780864915 Chr.22: 39600662-39600662 on Build GRCh38
rs1003000676 Chr.22: 39600567-39600567 on Build GRCh38
rs58365162 Chr.22: 39600667-39600667 on Build GRCh38
rs773108844 Chr.22: 39600529-39600529 on Build GRCh38
rs746503764 Chr.22: 39600656-39600656 on Build GRCh38
rs1024258859 Chr.22: 39600626-39600626 on Build GRCh38
rs764533152 Chr.22: 39600642-39600642 on Build GRCh38
rs12628240 Chr.22: 39600687-39600687 on Build GRCh38
rs766180233 Chr.22: 39600679-39600679 on Build GRCh38
rs868095512 Chr.22: 39600647-39600650 on Build GRCh38
rs754359024 Chr.22: 39600643-39600643 on Build GRCh38
rs759739015 Chr.22: 39600579-39600579 on Build GRCh38
rs370746198 Chr.22: 39600511-39600511 on Build GRCh38
rs771007686 Chr.22: 39600677-39600677 on Build GRCh38
rs60512481 Chr.22: 39600657-39600657 on Build GRCh38

View Full Product Details