Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58207759-58207998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
UniGene
Hs.509451
Gene Chromosome Location
Chr.14: 58200115-58235636 on Build GRCh38
Transcripts
NM_018477.2 XM_011536960.1 XM_011536961.1 XM_011536962.2 XM_011536963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5958183 c.154G>A p.V52I Chr.14: 58207939-58207939 on Build GRCh38
6463583 c.159A>G p.R53R Chr.14: 58207944-58207944 on Build GRCh38
6445946 c.151C>T p.P51S Chr.14: 58207936-58207936 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1016499591 Chr.14: 58207845-58207845 on Build GRCh38
rs972219116 Chr.14: 58207870-58207870 on Build GRCh38
rs777543813 Chr.14: 58207890-58207890 on Build GRCh38
rs376240327 Chr.14: 58207914-58207914 on Build GRCh38
rs991878417 Chr.14: 58207920-58207920 on Build GRCh38
rs767870589 Chr.14: 58207911-58207914 on Build GRCh38
rs1006654453 Chr.14: 58207814-58207814 on Build GRCh38
rs771683778 Chr.14: 58207933-58207933 on Build GRCh38
rs760399074 Chr.14: 58207941-58207941 on Build GRCh38
rs748974119 Chr.14: 58207899-58207899 on Build GRCh38
rs558397706 Chr.14: 58207903-58207903 on Build GRCh38
rs79640110 Chr.14: 58207932-58207932 on Build GRCh38
rs185801663 Chr.14: 58207809-58207809 on Build GRCh38
rs770777796 Chr.14: 58207901-58207901 on Build GRCh38
rs559393143 Chr.14: 58207957-58207957 on Build GRCh38
rs541086200 Chr.14: 58207936-58207936 on Build GRCh38
rs903273079 Chr.14: 58207876-58207876 on Build GRCh38
rs962638039 Chr.14: 58207860-58207860 on Build GRCh38
rs776437293 Chr.14: 58207964-58207964 on Build GRCh38
rs775196644 Chr.14: 58207939-58207939 on Build GRCh38

View Full Product Details