Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22100277-22100522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA3A
Entrez Gene ID
146177
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
-
UniGene
Hs.10697
Gene Chromosome Location
Chr.16: 22091930-22156966 on Build GRCh38
Transcripts
NM_173615.4 XM_011545742.2 XM_017022947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440916 c.352G>A p.E118K Chr.16: 22100417-22100417 on Build GRCh38
4387133 c.354G>A p.E118E Chr.16: 22100419-22100419 on Build GRCh38
968531 c.357G>A p.E119E Chr.16: 22100422-22100422 on Build GRCh38
3387287 c.341C>A p.T114K Chr.16: 22100309-22100309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372471067 Chr.16: 22100468-22100468 on Build GRCh38
rs891387058 Chr.16: 22100408-22100408 on Build GRCh38
rs377575921 Chr.16: 22100469-22100469 on Build GRCh38
rs765917274 Chr.16: 22100314-22100314 on Build GRCh38
rs534641937 Chr.16: 22100444-22100445 on Build GRCh38
rs369782153 Chr.16: 22100347-22100347 on Build GRCh38
rs902003598 Chr.16: 22100432-22100432 on Build GRCh38
rs757255650 Chr.16: 22100384-22100384 on Build GRCh38
rs72640484 Chr.16: 22100383-22100383 on Build GRCh38
rs756392828 Chr.16: 22100395-22100395 on Build GRCh38
rs1056103051 Chr.16: 22100329-22100329 on Build GRCh38
rs755814804 Chr.16: 22100374-22100374 on Build GRCh38
rs768927013 Chr.16: 22100397-22100397 on Build GRCh38
rs374019319 Chr.16: 22100337-22100337 on Build GRCh38
rs552730939 Chr.16: 22100340-22100340 on Build GRCh38
rs921333836 Chr.16: 22100382-22100382 on Build GRCh38
rs780257967 Chr.16: 22100441-22100441 on Build GRCh38
rs777764553 Chr.16: 22100459-22100459 on Build GRCh38
rs555388406 Chr.16: 22100481-22100481 on Build GRCh38
rs898179594 Chr.16: 22100447-22100447 on Build GRCh38
rs937180330 Chr.16: 22100324-22100324 on Build GRCh38
rs902853548 Chr.16: 22100352-22100352 on Build GRCh38
rs773543858 Chr.16: 22100301-22100301 on Build GRCh38
rs947037513 Chr.16: 22100339-22100339 on Build GRCh38
rs752563658 Chr.16: 22100372-22100372 on Build GRCh38
rs74345637 Chr.16: 22100303-22100303 on Build GRCh38
rs781232829 Chr.16: 22100416-22100416 on Build GRCh38
rs541691470 Chr.16: 22100435-22100435 on Build GRCh38

View Full Product Details