Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108991157-108991428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR47
Entrez Gene ID
22911
Gene Name
WD repeat domain 47
Gene Aliases
-
UniGene
Hs.570055
Gene Chromosome Location
Chr.1: 108970211-109042228 on Build GRCh38
Transcripts
NM_001142550.1 NM_001142551.1 NM_014969.5 XM_006710460.3 XM_011541027.2 XM_011541028.2 XM_011541029.1 XM_011541030.1 XM_011541031.1 XM_017000696.1 XM_017000697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6835223 c.1720C>T p.P574S Chr.1: 108991304-108991304 on Build GRCh38
7120733 c.1779G>T p.S593S Chr.1: 108991266-108991266 on Build GRCh38
7120734 c.1758G>T p.S586S Chr.1: 108991266-108991266 on Build GRCh38
6835222 c.1741C>T p.P581S Chr.1: 108991304-108991304 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761758544 Chr.1: 108991360-108991360 on Build GRCh38
rs922897082 Chr.1: 108991270-108991270 on Build GRCh38
rs763075968 Chr.1: 108991221-108991221 on Build GRCh38
rs747565723 Chr.1: 108991288-108991288 on Build GRCh38
rs754612572 Chr.1: 108991278-108991278 on Build GRCh38
rs774509199 Chr.1: 108991352-108991352 on Build GRCh38
rs780492881 Chr.1: 108991232-108991233 on Build GRCh38
rs768807635 Chr.1: 108991350-108991350 on Build GRCh38
rs753538948 Chr.1: 108991272-108991272 on Build GRCh38
rs761169120 Chr.1: 108991365-108991365 on Build GRCh38
rs933131663 Chr.1: 108991336-108991336 on Build GRCh38
rs767862406 Chr.1: 108991266-108991266 on Build GRCh38
rs752295707 Chr.1: 108991379-108991379 on Build GRCh38
rs375877843 Chr.1: 108991341-108991341 on Build GRCh38
rs773085936 Chr.1: 108991212-108991212 on Build GRCh38
rs768109050 Chr.1: 108991361-108991361 on Build GRCh38
rs140523650 Chr.1: 108991315-108991315 on Build GRCh38
rs770091041 Chr.1: 108991218-108991218 on Build GRCh38
rs933003171 Chr.1: 108991282-108991282 on Build GRCh38
rs374648649 Chr.1: 108991219-108991219 on Build GRCh38
rs772081855 Chr.1: 108991304-108991304 on Build GRCh38
rs368715513 Chr.1: 108991267-108991267 on Build GRCh38
rs777544291 Chr.1: 108991307-108991307 on Build GRCh38
rs762393246 Chr.1: 108991261-108991261 on Build GRCh38
rs368252643 Chr.1: 108991367-108991367 on Build GRCh38
rs779910866 Chr.1: 108991269-108991269 on Build GRCh38
rs139647836 Chr.1: 108991387-108991387 on Build GRCh38
rs751570294 Chr.1: 108991247-108991247 on Build GRCh38
rs1032435373 Chr.1: 108991250-108991250 on Build GRCh38
rs370990820 Chr.1: 108991285-108991285 on Build GRCh38
rs535172728 Chr.1: 108991193-108991193 on Build GRCh38
rs146364571 Chr.1: 108991377-108991377 on Build GRCh38
rs764151552 Chr.1: 108991245-108991245 on Build GRCh38
rs775753697 Chr.1: 108991324-108991324 on Build GRCh38
rs766857141 Chr.1: 108991366-108991366 on Build GRCh38
rs760350781 Chr.1: 108991217-108991217 on Build GRCh38
rs201032916 Chr.1: 108991375-108991375 on Build GRCh38

View Full Product Details