Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153506382-153506655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BGN
Entrez Gene ID
633
Gene Name
biglycan
Gene Aliases
DSPG1, PG-S1, PGI, SLRR1A
UniGene
Hs.821
Gene Chromosome Location
Chr.X: 153494889-153509554 on Build GRCh38
Transcripts
NM_001711.5 XM_017029724.1

SNP Detail

SNP ID Chromosome Location
rs782171412 Chr.X: 153506533-153506533 on Build GRCh38
rs782654370 Chr.X: 153506603-153506603 on Build GRCh38
rs782313100 Chr.X: 153506574-153506574 on Build GRCh38
rs782374654 Chr.X: 153506552-153506552 on Build GRCh38
rs782492275 Chr.X: 153506460-153506460 on Build GRCh38
rs1040804870 Chr.X: 153506504-153506504 on Build GRCh38
rs192912944 Chr.X: 153506443-153506443 on Build GRCh38
rs972221420 Chr.X: 153506435-153506435 on Build GRCh38
rs148808913 Chr.X: 153506536-153506536 on Build GRCh38
rs113726742 Chr.X: 153506527-153506527 on Build GRCh38
rs782083006 Chr.X: 153506579-153506579 on Build GRCh38
rs920805354 Chr.X: 153506438-153506438 on Build GRCh38
rs782643241 Chr.X: 153506503-153506503 on Build GRCh38
rs781871609 Chr.X: 153506521-153506521 on Build GRCh38
rs782730473 Chr.X: 153506484-153506484 on Build GRCh38
rs782797946 Chr.X: 153506431-153506431 on Build GRCh38
rs781822305 Chr.X: 153506489-153506489 on Build GRCh38
rs782737792 Chr.X: 153506599-153506599 on Build GRCh38
rs782679609 Chr.X: 153506524-153506524 on Build GRCh38
rs910888540 Chr.X: 153506453-153506453 on Build GRCh38
rs952335300 Chr.X: 153506439-153506439 on Build GRCh38
rs782001829 Chr.X: 153506555-153506555 on Build GRCh38
rs782231842 Chr.X: 153506541-153506541 on Build GRCh38
rs11538655 Chr.X: 153506581-153506581 on Build GRCh38
rs781869020 Chr.X: 153506635-153506635 on Build GRCh38
rs781790595 Chr.X: 153506567-153506567 on Build GRCh38
rs935930397 Chr.X: 153506578-153506578 on Build GRCh38
rs782553600 Chr.X: 153506593-153506593 on Build GRCh38
rs1024372254 Chr.X: 153506420-153506420 on Build GRCh38
rs781882458 Chr.X: 153506505-153506505 on Build GRCh38
rs375019160 Chr.X: 153506513-153506513 on Build GRCh38
rs367995322 Chr.X: 153506575-153506575 on Build GRCh38
rs782602954 Chr.X: 153506537-153506537 on Build GRCh38
rs782574567 Chr.X: 153506493-153506494 on Build GRCh38
rs371729412 Chr.X: 153506493-153506493 on Build GRCh38

View Full Product Details