Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53380557-53380802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC1A
Entrez Gene ID
8243
Gene Name
structural maintenance of chromosomes 1A
Gene Aliases
CDLS2, DXS423E, SB1.8, SMC1, SMC1L1, SMC1alpha, SMCB
UniGene
Hs.211602
Gene Chromosome Location
Chr.X: 53374149-53422728 on Build GRCh38
Transcripts
NM_001281463.1 NM_006306.3

SNP Detail

SNP ID Chromosome Location
rs146216425 Chr.X: 53380647-53380647 on Build GRCh38
rs782673881 Chr.X: 53380759-53380759 on Build GRCh38
rs782534679 Chr.X: 53380744-53380744 on Build GRCh38
rs782055278 Chr.X: 53380654-53380654 on Build GRCh38
rs966209670 Chr.X: 53380762-53380762 on Build GRCh38
rs781998306 Chr.X: 53380627-53380627 on Build GRCh38
rs781825792 Chr.X: 53380671-53380671 on Build GRCh38
rs781785288 Chr.X: 53380750-53380750 on Build GRCh38
rs782683440 Chr.X: 53380607-53380607 on Build GRCh38
rs869312954 Chr.X: 53380660-53380660 on Build GRCh38
rs782319907 Chr.X: 53380612-53380612 on Build GRCh38
rs782252473 Chr.X: 53380765-53380765 on Build GRCh38
rs782504122 Chr.X: 53380583-53380583 on Build GRCh38
rs782361939 Chr.X: 53380770-53380770 on Build GRCh38
rs863225459 Chr.X: 53380685-53380686 on Build GRCh38
rs587784419 Chr.X: 53380681-53380681 on Build GRCh38
rs1033021951 Chr.X: 53380685-53380685 on Build GRCh38
rs181086310 Chr.X: 53380755-53380755 on Build GRCh38
rs782267492 Chr.X: 53380613-53380613 on Build GRCh38
rs782763816 Chr.X: 53380704-53380704 on Build GRCh38
rs956449031 Chr.X: 53380735-53380735 on Build GRCh38
rs782493076 Chr.X: 53380753-53380753 on Build GRCh38
rs782581458 Chr.X: 53380745-53380745 on Build GRCh38
rs782278656 Chr.X: 53380610-53380610 on Build GRCh38
rs781943084 Chr.X: 53380653-53380653 on Build GRCh38
rs782336810 Chr.X: 53380616-53380616 on Build GRCh38
rs782175064 Chr.X: 53380646-53380646 on Build GRCh38
rs138015913 Chr.X: 53380668-53380668 on Build GRCh38
rs782128162 Chr.X: 53380680-53380680 on Build GRCh38
rs782220435 Chr.X: 53380614-53380614 on Build GRCh38

View Full Product Details