Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17453548-17453811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST8SIA6
Entrez Gene ID
338596
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Gene Aliases
SIA8F, SIAT8F, ST8SIA-VI
UniGene
Hs.677766
Gene Chromosome Location
Chr.10: 17315414-17455966 on Build GRCh38
Transcripts
NM_001004470.1 XM_017016174.1 XM_017016175.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743385 c.148G>A p.A50T Chr.10: 17453611-17453611 on Build GRCh38
6129037 c.197A>T p.N66I Chr.10: 17453562-17453562 on Build GRCh38
6129036 c.164G>A p.R55Q Chr.10: 17453595-17453595 on Build GRCh38
7352435 c.139A>T p.T47S Chr.10: 17453620-17453620 on Build GRCh38
3985541 c.150G>A p.A50A Chr.10: 17453609-17453609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532150579 Chr.10: 17453639-17453639 on Build GRCh38
rs750181246 Chr.10: 17453652-17453652 on Build GRCh38
rs928575756 Chr.10: 17453570-17453570 on Build GRCh38
rs778490508 Chr.10: 17453600-17453600 on Build GRCh38
rs775563615 Chr.10: 17453623-17453623 on Build GRCh38
rs954812272 Chr.10: 17453692-17453692 on Build GRCh38
rs757732772 Chr.10: 17453699-17453699 on Build GRCh38
rs886741369 Chr.10: 17453593-17453593 on Build GRCh38
rs767244447 Chr.10: 17453635-17453635 on Build GRCh38
rs748747905 Chr.10: 17453596-17453596 on Build GRCh38
rs369560766 Chr.10: 17453624-17453624 on Build GRCh38
rs752054269 Chr.10: 17453698-17453698 on Build GRCh38
rs1014376281 Chr.10: 17453580-17453580 on Build GRCh38
rs370019838 Chr.10: 17453694-17453694 on Build GRCh38
rs925847780 Chr.10: 17453742-17453742 on Build GRCh38
rs374099274 Chr.10: 17453695-17453695 on Build GRCh38
rs754576924 Chr.10: 17453602-17453602 on Build GRCh38
rs569216942 Chr.10: 17453730-17453730 on Build GRCh38
rs766061091 Chr.10: 17453678-17453678 on Build GRCh38
rs45462798 Chr.10: 17453620-17453620 on Build GRCh38
rs963545056 Chr.10: 17453776-17453776 on Build GRCh38
rs372934682 Chr.10: 17453597-17453597 on Build GRCh38
rs751826430 Chr.10: 17453655-17453656 on Build GRCh38
rs934459481 Chr.10: 17453598-17453598 on Build GRCh38
rs761774544 Chr.10: 17453632-17453632 on Build GRCh38
rs781439649 Chr.10: 17453707-17453707 on Build GRCh38
rs746109264 Chr.10: 17453619-17453619 on Build GRCh38
rs1052415364 Chr.10: 17453725-17453725 on Build GRCh38
rs774335113 Chr.10: 17453631-17453631 on Build GRCh38
rs45551937 Chr.10: 17453710-17453710 on Build GRCh38
rs760256547 Chr.10: 17453669-17453669 on Build GRCh38
rs1025256981 Chr.10: 17453588-17453588 on Build GRCh38
rs978689676 Chr.10: 17453735-17453735 on Build GRCh38
rs771249416 Chr.10: 17453609-17453609 on Build GRCh38
rs753535099 Chr.10: 17453680-17453680 on Build GRCh38
rs778216377 Chr.10: 17453611-17453611 on Build GRCh38
rs1055630130 Chr.10: 17453755-17453755 on Build GRCh38
rs367584455 Chr.10: 17453601-17453601 on Build GRCh38
rs937195849 Chr.10: 17453743-17453743 on Build GRCh38
rs779451125 Chr.10: 17453586-17453586 on Build GRCh38
rs755749073 Chr.10: 17453566-17453566 on Build GRCh38
rs554515980 Chr.10: 17453773-17453773 on Build GRCh38
rs947106753 Chr.10: 17453655-17453655 on Build GRCh38
rs143249700 Chr.10: 17453765-17453765 on Build GRCh38
rs781589985 Chr.10: 17453612-17453612 on Build GRCh38
rs1016416369 Chr.10: 17453766-17453766 on Build GRCh38
rs1029513191 Chr.10: 17453671-17453671 on Build GRCh38
rs1016446369 Chr.10: 17453617-17453617 on Build GRCh38

View Full Product Details