Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51908635-51908908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMOD3
Entrez Gene ID
29766
Gene Name
tropomodulin 3
Gene Aliases
UTMOD
UniGene
Hs.4998
Gene Chromosome Location
Chr.15: 51829628-51912138 on Build GRCh38
Transcripts
NM_014547.4 XM_017022087.1 XM_017022088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6538302 c.1031A>G p.K344R Chr.15: 51908782-51908782 on Build GRCh38
6804486 c.1026G>A p.V342V Chr.15: 51908777-51908777 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758413403 Chr.15: 51908773-51908773 on Build GRCh38
rs776441187 Chr.15: 51908779-51908780 on Build GRCh38
rs547912315 Chr.15: 51908833-51908833 on Build GRCh38
rs773203028 Chr.15: 51908677-51908678 on Build GRCh38
rs1001794014 Chr.15: 51908681-51908681 on Build GRCh38
rs779060752 Chr.15: 51908857-51908857 on Build GRCh38
rs750587148 Chr.15: 51908772-51908772 on Build GRCh38
rs772756242 Chr.15: 51908734-51908734 on Build GRCh38
rs770343233 Chr.15: 51908822-51908822 on Build GRCh38
rs529994559 Chr.15: 51908823-51908823 on Build GRCh38
rs762355388 Chr.15: 51908740-51908740 on Build GRCh38
rs372092293 Chr.15: 51908867-51908867 on Build GRCh38
rs78819931 Chr.15: 51908743-51908743 on Build GRCh38
rs780124953 Chr.15: 51908778-51908778 on Build GRCh38
rs759290865 Chr.15: 51908841-51908841 on Build GRCh38
rs200837938 Chr.15: 51908837-51908837 on Build GRCh38
rs1045442744 Chr.15: 51908666-51908666 on Build GRCh38
rs747220326 Chr.15: 51908779-51908779 on Build GRCh38
rs752511603 Chr.15: 51908752-51908752 on Build GRCh38
rs199520707 Chr.15: 51908797-51908797 on Build GRCh38
rs1034483513 Chr.15: 51908699-51908699 on Build GRCh38
rs764117440 Chr.15: 51908771-51908771 on Build GRCh38
rs148821059 Chr.15: 51908794-51908794 on Build GRCh38
rs748489254 Chr.15: 51908806-51908806 on Build GRCh38
rs530504000 Chr.15: 51908750-51908750 on Build GRCh38
rs925391614 Chr.15: 51908712-51908712 on Build GRCh38
rs993856956 Chr.15: 51908665-51908665 on Build GRCh38
rs543893161 Chr.15: 51908704-51908704 on Build GRCh38
rs888235768 Chr.15: 51908872-51908872 on Build GRCh38
rs374970124 Chr.15: 51908827-51908827 on Build GRCh38
rs202127944 Chr.15: 51908741-51908741 on Build GRCh38
rs907424953 Chr.15: 51908671-51908671 on Build GRCh38
rs768583599 Chr.15: 51908732-51908733 on Build GRCh38
rs757270688 Chr.15: 51908787-51908787 on Build GRCh38
rs763018645 Chr.15: 51908849-51908850 on Build GRCh38
rs976032846 Chr.15: 51908814-51908814 on Build GRCh38
rs756172786 Chr.15: 51908770-51908770 on Build GRCh38
rs773859023 Chr.15: 51908828-51908828 on Build GRCh38
rs372693419 Chr.15: 51908751-51908751 on Build GRCh38
rs749732321 Chr.15: 51908825-51908825 on Build GRCh38
rs569625038 Chr.15: 51908875-51908875 on Build GRCh38
rs369394086 Chr.15: 51908659-51908659 on Build GRCh38

View Full Product Details