Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85654114-85654359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM126A
Entrez Gene ID
84233
Gene Name
transmembrane protein 126A
Gene Aliases
OPA7
UniGene
Hs.533725
Gene Chromosome Location
Chr.11: 85647919-85656553 on Build GRCh38
Transcripts
NM_001244735.1 NM_032273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
76750 c.179T>C p.V60A Chr.11: 85654155-85654155 on Build GRCh38
5902994 c.245C>G p.T82S Chr.11: 85654221-85654221 on Build GRCh38
3810572 c.273G>T p.L91F Chr.11: 85654249-85654249 on Build GRCh38
5018860 c.225delT p.T78fs*4 Chr.11: 85654201-85654201 on Build GRCh38
4037185 c.163C>T p.R55* Chr.11: 85654139-85654139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777641763 Chr.11: 85654237-85654237 on Build GRCh38
rs11556797 Chr.11: 85654167-85654167 on Build GRCh38
rs780975544 Chr.11: 85654255-85654255 on Build GRCh38
rs148446292 Chr.11: 85654187-85654187 on Build GRCh38
rs759862219 Chr.11: 85654289-85654289 on Build GRCh38
rs1042977900 Chr.11: 85654247-85654247 on Build GRCh38
rs761278708 Chr.11: 85654275-85654275 on Build GRCh38
rs904221195 Chr.11: 85654268-85654268 on Build GRCh38
rs777049279 Chr.11: 85654282-85654282 on Build GRCh38
rs565261459 Chr.11: 85654209-85654209 on Build GRCh38
rs776223013 Chr.11: 85654158-85654158 on Build GRCh38
rs771624394 Chr.11: 85654280-85654280 on Build GRCh38
rs769482743 Chr.11: 85654263-85654263 on Build GRCh38
rs745704252 Chr.11: 85654258-85654258 on Build GRCh38
rs561429591 Chr.11: 85654152-85654152 on Build GRCh38
rs768104829 Chr.11: 85654171-85654171 on Build GRCh38
rs766857004 Chr.11: 85654207-85654207 on Build GRCh38
rs377476349 Chr.11: 85654166-85654166 on Build GRCh38
rs762733239 Chr.11: 85654234-85654235 on Build GRCh38
rs1032026676 Chr.11: 85654330-85654330 on Build GRCh38
rs377748343 Chr.11: 85654179-85654179 on Build GRCh38
rs532521030 Chr.11: 85654216-85654216 on Build GRCh38
rs200692120 Chr.11: 85654256-85654256 on Build GRCh38
rs900924118 Chr.11: 85654328-85654328 on Build GRCh38
rs888552081 Chr.11: 85654140-85654140 on Build GRCh38
rs371334632 Chr.11: 85654210-85654210 on Build GRCh38
rs768885199 Chr.11: 85654239-85654239 on Build GRCh38
rs548089203 Chr.11: 85654301-85654301 on Build GRCh38
rs745535321 Chr.11: 85654161-85654161 on Build GRCh38
rs192269199 Chr.11: 85654143-85654143 on Build GRCh38
rs560111095 Chr.11: 85654328-85654328 on Build GRCh38
rs1026249707 Chr.11: 85654332-85654332 on Build GRCh38
rs536867858 Chr.11: 85654317-85654317 on Build GRCh38
rs770737681 Chr.11: 85654157-85654157 on Build GRCh38
rs768302604 Chr.11: 85654273-85654273 on Build GRCh38
rs569477121 Chr.11: 85654314-85654314 on Build GRCh38
rs765042337 Chr.11: 85654264-85654264 on Build GRCh38
rs121434508 Chr.11: 85654139-85654139 on Build GRCh38
rs765478259 Chr.11: 85654290-85654290 on Build GRCh38
rs1017898672 Chr.11: 85654279-85654279 on Build GRCh38
rs755215136 Chr.11: 85654217-85654217 on Build GRCh38
rs149739249 Chr.11: 85654142-85654142 on Build GRCh38
rs745619852 Chr.11: 85654217-85654232 on Build GRCh38
rs529891145 Chr.11: 85654299-85654299 on Build GRCh38
rs766191226 Chr.11: 85654280-85654280 on Build GRCh38
rs112880915 Chr.11: 85654331-85654331 on Build GRCh38
rs762436987 Chr.11: 85654170-85654170 on Build GRCh38
rs761082602 Chr.11: 85654194-85654194 on Build GRCh38
rs865878230 Chr.11: 85654150-85654150 on Build GRCh38
rs547511651 Chr.11: 85654250-85654250 on Build GRCh38
rs774073139 Chr.11: 85654296-85654297 on Build GRCh38
rs142626884 Chr.11: 85654235-85654235 on Build GRCh38
rs117453673 Chr.11: 85654270-85654270 on Build GRCh38

View Full Product Details