Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21962357-21962614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD9
Entrez Gene ID
255762
Gene Name
PDZ domain containing 9
Gene Aliases
C16orf65
UniGene
Hs.98244
Gene Chromosome Location
Chr.16: 21957540-22001137 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Gene Symbol
UQCRC2
Entrez Gene ID
7385
Gene Name
ubiquinol-cytochrome c reductase core protein II
Gene Aliases
MC3DN5, QCR2, UQCR2
UniGene
Hs.528803
Gene Chromosome Location
Chr.16: 21953064-21983660 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6817717 c.333-8C>T p.? Chr.16: 21962452-21962452 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs915998978 Chr.16: 21962549-21962549 on Build GRCh38
rs139040421 Chr.16: 21962453-21962453 on Build GRCh38
rs369630012 Chr.16: 21962466-21962466 on Build GRCh38
rs771836280 Chr.16: 21962501-21962501 on Build GRCh38
rs751927180 Chr.16: 21962432-21962432 on Build GRCh38
rs745667374 Chr.16: 21962491-21962491 on Build GRCh38
rs146146335 Chr.16: 21962440-21962440 on Build GRCh38
rs766822832 Chr.16: 21962389-21962389 on Build GRCh38
rs763111546 Chr.16: 21962534-21962534 on Build GRCh38
rs138177262 Chr.16: 21962467-21962467 on Build GRCh38
rs904383898 Chr.16: 21962591-21962591 on Build GRCh38
rs567343943 Chr.16: 21962533-21962533 on Build GRCh38
rs752775175 Chr.16: 21962577-21962577 on Build GRCh38
rs894577937 Chr.16: 21962459-21962459 on Build GRCh38
rs766678249 Chr.16: 21962431-21962431 on Build GRCh38
rs376496884 Chr.16: 21962452-21962452 on Build GRCh38
rs778618710 Chr.16: 21962488-21962488 on Build GRCh38
rs143434256 Chr.16: 21962507-21962507 on Build GRCh38
rs373451853 Chr.16: 21962515-21962515 on Build GRCh38
rs1028065038 Chr.16: 21962426-21962426 on Build GRCh38
rs994877543 Chr.16: 21962446-21962446 on Build GRCh38
rs753205581 Chr.16: 21962451-21962451 on Build GRCh38
rs368690692 Chr.16: 21962465-21962465 on Build GRCh38
rs778437483 Chr.16: 21962454-21962455 on Build GRCh38
rs897790044 Chr.16: 21962395-21962395 on Build GRCh38
rs779889317 Chr.16: 21962506-21962506 on Build GRCh38
rs759885865 Chr.16: 21962439-21962439 on Build GRCh38
rs986332281 Chr.16: 21962458-21962458 on Build GRCh38
rs117356220 Chr.16: 21962573-21962573 on Build GRCh38
rs543510412 Chr.16: 21962492-21962492 on Build GRCh38
rs972331534 Chr.16: 21962490-21962490 on Build GRCh38
rs753310135 Chr.16: 21962461-21962461 on Build GRCh38
rs141608385 Chr.16: 21962563-21962563 on Build GRCh38
rs561278357 Chr.16: 21962576-21962576 on Build GRCh38
rs768692859 Chr.16: 21962512-21962512 on Build GRCh38
rs763317327 Chr.16: 21962427-21962427 on Build GRCh38

View Full Product Details