Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146469601-146469862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCERG1
Entrez Gene ID
10915
Gene Name
transcription elongation regulator 1
Gene Aliases
CA150, TAF2S, Urn1
UniGene
Hs.443465
Gene Chromosome Location
Chr.5: 146447124-146511508 on Build GRCh38
Transcripts
NM_001040006.1 NM_006706.3 XM_005268365.2 XM_017008977.1 XM_017008978.1 XM_017008979.1 XM_017008980.1 XM_017008981.1 XM_017008982.1 XM_017008983.1 XM_017008984.1 XM_017008985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1672012 c.1393G>A p.E465K Chr.5: 146469738-146469738 on Build GRCh38
5867735 c.1322C>T p.A441V Chr.5: 146469667-146469667 on Build GRCh38
1329003 c.1260A>G p.A420A Chr.5: 146469605-146469605 on Build GRCh38
5060816 c.1282G>A p.G428R Chr.5: 146469627-146469627 on Build GRCh38
6798203 c.1343_1345delATA p.N450delN Chr.5: 146469688-146469690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770309172 Chr.5: 146469625-146469625 on Build GRCh38
rs761070448 Chr.5: 146469689-146469689 on Build GRCh38
rs780459218 Chr.5: 146469783-146469783 on Build GRCh38
rs754302262 Chr.5: 146469755-146469755 on Build GRCh38
rs376401078 Chr.5: 146469642-146469642 on Build GRCh38
rs201116640 Chr.5: 146469768-146469768 on Build GRCh38
rs139746932 Chr.5: 146469712-146469712 on Build GRCh38
rs954115152 Chr.5: 146469713-146469713 on Build GRCh38
rs267600471 Chr.5: 146469733-146469733 on Build GRCh38
rs764008265 Chr.5: 146469768-146469769 on Build GRCh38
rs758542077 Chr.5: 146469782-146469782 on Build GRCh38
rs546139607 Chr.5: 146469813-146469813 on Build GRCh38
rs921731235 Chr.5: 146469652-146469652 on Build GRCh38
rs764564234 Chr.5: 146469722-146469722 on Build GRCh38
rs920452020 Chr.5: 146469807-146469807 on Build GRCh38
rs201358092 Chr.5: 146469759-146469759 on Build GRCh38
rs929123240 Chr.5: 146469812-146469812 on Build GRCh38
rs200743119 Chr.5: 146469761-146469761 on Build GRCh38
rs751325514 Chr.5: 146469784-146469785 on Build GRCh38
rs1053105727 Chr.5: 146469801-146469801 on Build GRCh38
rs973334896 Chr.5: 146469800-146469800 on Build GRCh38
rs267600470 Chr.5: 146469718-146469718 on Build GRCh38
rs549098268 Chr.5: 146469810-146469810 on Build GRCh38
rs773920453 Chr.5: 146469629-146469629 on Build GRCh38
rs775829176 Chr.5: 146469674-146469674 on Build GRCh38
rs913514022 Chr.5: 146469814-146469814 on Build GRCh38
rs191593593 Chr.5: 146469656-146469656 on Build GRCh38
rs755922119 Chr.5: 146469794-146469794 on Build GRCh38
rs201021166 Chr.5: 146469787-146469787 on Build GRCh38

View Full Product Details