Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32650223-32650496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf203
Entrez Gene ID
284805
Gene Name
chromosome 20 open reading frame 203
Gene Aliases
-
UniGene
Hs.353262
Gene Chromosome Location
Chr.20: 32631625-32651981 on Build GRCh38
Transcripts
NM_182584.2

SNP Detail

SNP ID Chromosome Location
rs972768119 Chr.20: 32650283-32650283 on Build GRCh38
rs1052557416 Chr.20: 32650358-32650358 on Build GRCh38
rs1026079210 Chr.20: 32650444-32650444 on Build GRCh38
rs541360415 Chr.20: 32650301-32650301 on Build GRCh38
rs958703122 Chr.20: 32650399-32650399 on Build GRCh38
rs531439283 Chr.20: 32650419-32650419 on Build GRCh38
rs942303113 Chr.20: 32650265-32650265 on Build GRCh38
rs889965485 Chr.20: 32650418-32650418 on Build GRCh38
rs773987728 Chr.20: 32650393-32650393 on Build GRCh38
rs1043465662 Chr.20: 32650326-32650326 on Build GRCh38
rs890783164 Chr.20: 32650251-32650251 on Build GRCh38
rs918577861 Chr.20: 32650288-32650288 on Build GRCh38
rs187868533 Chr.20: 32650308-32650308 on Build GRCh38
rs994239901 Chr.20: 32650443-32650444 on Build GRCh38
rs761370020 Chr.20: 32650400-32650400 on Build GRCh38
rs771816188 Chr.20: 32650408-32650408 on Build GRCh38
rs925069457 Chr.20: 32650330-32650330 on Build GRCh38
rs764433662 Chr.20: 32650442-32650442 on Build GRCh38
rs772578690 Chr.20: 32650412-32650412 on Build GRCh38
rs992489240 Chr.20: 32650449-32650449 on Build GRCh38
rs901477966 Chr.20: 32650367-32650367 on Build GRCh38
rs17123507 Chr.20: 32650282-32650282 on Build GRCh38
rs760220578 Chr.20: 32650413-32650413 on Build GRCh38
rs956521471 Chr.20: 32650448-32650448 on Build GRCh38
rs35103329 Chr.20: 32650337-32650337 on Build GRCh38
rs374459215 Chr.20: 32650424-32650424 on Build GRCh38
rs542460541 Chr.20: 32650319-32650319 on Build GRCh38
rs752015993 Chr.20: 32650471-32650471 on Build GRCh38
rs183179301 Chr.20: 32650302-32650302 on Build GRCh38
rs1039651741 Chr.20: 32650268-32650268 on Build GRCh38
rs947852642 Chr.20: 32650322-32650322 on Build GRCh38
rs753228595 Chr.20: 32650429-32650429 on Build GRCh38
rs764271074 Chr.20: 32650306-32650306 on Build GRCh38
rs763232893 Chr.20: 32650430-32650430 on Build GRCh38
rs1051312933 Chr.20: 32650406-32650407 on Build GRCh38
rs1035735233 Chr.20: 32650363-32650363 on Build GRCh38
rs571146882 Chr.20: 32650445-32650445 on Build GRCh38

View Full Product Details